Assay Details
| Gene Symbol: KCNH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204798.1 | NP_001191727.1 | 1 - 2 | 436 | 82 | |
| NM_172057.2 | NP_742054.1 | 1 - 2 | 436 | 82 | ||
| GenBank mRNA | AJ512214.1 | - | 1 - 2 | 127 | 82 | |
| AJ609614.1 | - | 1 - 2 | 110 | 82 | ||
| BG683210.1 | - | 1 - 2 | 435 | 82 | ||
| BQ890248.1 | - | 1 - 2 | 434 | 82 | ||
| DQ120125.1 | - | 1 - 2 | 513 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07002024.hg.1
Set Membership: |
|

KCNH2
Genomic Map