Assay Details
| Gene Symbol: CLVS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173519.2 | NP_775790.1 | 2 - 3 | 773 | 76 | |
| XM_017013141.1 | XP_016868630.1 | 3 - 4 | 764 | 76 | ||
| XM_017013142.1 | XP_016868631.1 | 3 - 4 | 921 | 76 | ||
| GenBank mRNA | AF445194.1 | - | 2 - 3 | 767 | 76 | |
| AK313040.1 | - | 2 - 3 | 720 | 76 | ||
| AY094971.1 | - | 2 - 3 | 731 | 76 | ||
| BC042617.1 | - | 2 - 3 | 748 | 5192332 | 76 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000414.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08005810.hg.1 | TC08000414.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005810.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005810.hg.1 | ENST00000519846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005810.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000518426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000518426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000518426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000518592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000518592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000518592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000518592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000518592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000518592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000518592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000518592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000518592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000518858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000518858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000518858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000518858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000520712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000520712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000520712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000520712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000520712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000520712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000521189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000521189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000521189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000521189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000521189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000521189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000521189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000522621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000522621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000522621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000522621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000522621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000522928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000522928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000522928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000522928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000523868.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000523868.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000523868.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000523868.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000523868.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000524095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000524095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000524095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | ENST00000524095.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005814.hg.1 | TC08000414.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005814.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005814.hg.1 | ENST00000518592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005814.hg.1 | ENST00000519846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005814.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000518426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000518426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000518426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000518592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000518592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000518592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000518592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000518592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000518592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000518592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000518592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000518592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000518858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000518858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000518858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000518858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000520712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000520712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000520712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000520712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000520712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000520712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000521189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000521189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000521189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000521189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000521189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000521189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000521189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000522621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000522621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000522621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000522621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000522621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000522928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000522928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000522928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000522928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000523868.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000523868.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000523868.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000523868.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000523868.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000524095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000524095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000524095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | ENST00000524095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007787.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800153559.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153559.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153559.hg.1 | ENST00000521189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153559.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800153559.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153559.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153559.hg.1 | ENST00000521189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153559.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800153559.hg.1 | TC0800007787.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800153563.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153563.hg.1 | ENST00000518592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153563.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153563.hg.1 | ENST00000520712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153563.hg.1 | ENST00000521189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153563.hg.1 | ENST00000524095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153563.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800153563.hg.1 | ENST00000325897.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153563.hg.1 | ENST00000518592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153563.hg.1 | ENST00000519846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153563.hg.1 | ENST00000520712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153563.hg.1 | ENST00000521189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153563.hg.1 | ENST00000524095.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153563.hg.1 | NM_173519.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800153563.hg.1 | TC0800007787.hg.1 | Clariom D TC |

CLVS1
Genomic Map