Assay Details
Gene Symbol: C9orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173520.2 | NP_775791.1 | 2 - 3 | 657 | 78 | |
GenBank mRNA | BC034752.1 | - | 2 - 3 | 657 | 5193354 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003908 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003838 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003844 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353561 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353588 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003774 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003805 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003882 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003826 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16274647 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003830 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003786 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003884 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353586 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353465 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353494 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003876 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16274650 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003900 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003840 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003862 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003887 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353550 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003910 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003827 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003868 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003875 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003914 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003777 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353480 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353611 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003863 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003811 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003822 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16274644 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003785 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003821 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003925 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003818 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003858 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353506 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003861 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003924 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003810 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003864 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003920 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003907 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353575 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003919 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003902 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003833 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003903 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003799 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16274656 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353590 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353500 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003809 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353571 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353580 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003848 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003897 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353477 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353562 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353459 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003871 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353538 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003781 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353560 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003765 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353616 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353594 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003802 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003801 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353504 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353513 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003808 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353582 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003873 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003819 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003847 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003800 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003771 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003886 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003835 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353468 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003829 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003921 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353567 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003768 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003775 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003803 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003789 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003850 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003832 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003878 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003823 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353587 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003874 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003870 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003814 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003788 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353606 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003782 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353573 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003879 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003849 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353542 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16274659 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003856 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003795 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003905 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003894 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353572 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003807 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353456 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353488 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003881 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003865 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003891 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003859 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353529 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003767 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353517 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003796 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003790 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003916 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003791 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003784 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353483 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353484 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003854 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353555 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003911 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003857 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27353539 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003841 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003824 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P303874 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 9277 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P120080 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P202119 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P120080 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 4502 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P303874 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P120080 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P202119 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P202119 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P120080 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 87868 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 45624 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 8909 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 80274 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P202119 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P120080 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P32574614 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P433138 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8159241 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8159242 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8159240 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8159239 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1553919_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0900009155.hg.1 | ENST00000623103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009155.hg.1 | ENST00000623103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009155.hg.1 | ENST00000623103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009155.hg.1 | NM_173520.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009155.hg.1 | NM_173520.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009155.hg.1 | NM_173520.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900162335.hg.1 | ENST00000623103.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900162335.hg.1 | NM_173520.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900162335.hg.1 | ENST00000623103.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900162335.hg.1 | NM_173520.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900162335.hg.1 | TC0900009155.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900009155.hg.1 | ENST00000623103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009155.hg.1 | ENST00000623103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009155.hg.1 | ENST00000623103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009155.hg.1 | NM_173520.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009155.hg.1 | NM_173520.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009155.hg.1 | NM_173520.2 | Refseq |
Human Genome U133 Plus 2.0 Array | 1553919_at | - | - |
Human Genome U133 Plus 2.0 Array | 1553919_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553919_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553919_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553919_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553919_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553919_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553919_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553919_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553919_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553919_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553919_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553919_at3 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 225668 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1815635 | BC034752.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1815635 | BC034752.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1815635 | BC034752.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1815635 | BC034752.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1815635 | BC034752.1 | human_transcriptome |
