Assay Details
Gene Symbol: PRR14L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173566.2 | NP_775837.2 | 5 - 6 | 6021 | 73 | |
XM_006724219.3 | XP_006724282.1 | 5 - 6 | 6581 | 73 | ||
XM_006724220.3 | XP_006724283.1 | 5 - 6 | 5983 | 73 | ||
XM_006724221.2 | XP_006724284.1 | 4 - 5 | 5537 | 73 | ||
XM_011530080.2 | XP_011528382.1 | 6 - 7 | 6789 | 73 | ||
XM_011530081.2 | XP_011528383.1 | 5 - 6 | 6665 | 73 | ||
XM_017028736.1 | XP_016884225.1 | 5 - 6 | 7741 | 73 | ||
XM_017028737.1 | XP_016884226.1 | 4 - 5 | 7179 | 73 | ||
GenBank mRNA | AK130944.1 | - | 2 - 3 | 1835 | 73 | |
BC040859.1 | - | 2 - 3 | 735 | 5535580 | 73 | |
BX647272.1 | - | 2 - 3 | 324 | 73 | ||
CT841517.1 | - | 4 - 5 | 5853 | 73 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22001490.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR22011933.hg.1 | TC22001490.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22011933.hg.1 | ENST00000327423.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22011933.hg.1 | ENST00000397493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22011933.hg.1 | ENST00000434485.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22011933.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200161148.hg.1 | TC2200008489.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200161148.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200161148.hg.1 | ENST00000330495.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200161148.hg.1 | ENST00000431684.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200161148.hg.1 | ENST00000432485.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200161148.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200161148.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200161148.hg.1 | ENST00000330495.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200161148.hg.1 | ENST00000431684.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200161148.hg.1 | ENST00000432485.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200161148.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
