Assay Details
Gene Symbol: ZNF663P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL831934.1 | - | 1 - 2 | 62 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AL831934.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839360 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191402 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191406 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191351 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191367 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839439 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191393 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17195698 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191295 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839421 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839500 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191433 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191357 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839462 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839527 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839485 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191356 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839468 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17195711 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191301 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191365 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191428 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839446 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191401 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839364 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839481 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839437 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191408 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839528 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191429 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839440 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191431 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191416 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839441 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839530 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191409 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839493 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191438 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191410 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17195717 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839498 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191297 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191394 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17195716 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191359 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191449 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839522 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839482 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839491 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839426 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839461 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191298 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839442 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839445 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191362 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839449 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839489 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17195704 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839448 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191358 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191403 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17195701 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839427 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191422 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191437 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191442 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839469 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839356 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839450 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191445 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839516 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839435 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839519 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191427 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839362 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191448 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191350 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839466 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191392 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839447 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839480 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839520 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191399 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839350 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839525 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839463 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839483 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839464 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839418 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839531 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839526 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839357 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839502 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839521 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31839488 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191376 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11191370 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P44992 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3208995 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 1689 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P117299 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P103300 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P117299 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P103300 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 16507 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 47353 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P103300 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P117299 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P03525241 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P21144998 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03525241 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P117299 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P103300 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P44992 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 10139 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 162922 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 76700 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 179594 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 78980 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 87212 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 72280 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03525250 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P117299 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P21144994 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P21144998 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P21144953 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P13808775 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P41339893 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03525282 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03525265 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P13811125 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P103300 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P21144981 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P21144966 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03525247 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03525241 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P41339862 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P41339871 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P21145007 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P21144957 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P911217 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8066740 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8066743 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8066742 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8066744 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8066741 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8066739 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1553911_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC2000009270.hg.1 | ENST00000400371.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009270.hg.1 | ENST00000400371.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009270.hg.1 | ENST00000400371.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009270.hg.1 | ENST00000400371.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009270.hg.1 | ENST00000400371.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009270.hg.1 | ENST00000400371.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009270.hg.1 | ENST00000400371.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009270.hg.1 | ENST00000400371.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009270.hg.1 | ENST00000400371.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009270.hg.1 | ENST00000472805.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009270.hg.1 | ENST00000472805.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009270.hg.1 | ENST00000472805.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009270.hg.1 | ENST00000472805.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009270.hg.1 | NR_045983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009270.hg.1 | NR_045983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009270.hg.1 | NR_045983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009270.hg.1 | NR_045983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009270.hg.1 | NR_045983.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009270.hg.1 | NR_045983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009270.hg.1 | ENST00000400371.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009270.hg.1 | ENST00000400371.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009270.hg.1 | ENST00000400371.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009270.hg.1 | ENST00000400371.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009270.hg.1 | ENST00000400371.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009270.hg.1 | ENST00000400371.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009270.hg.1 | ENST00000400371.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009270.hg.1 | ENST00000400371.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009270.hg.1 | ENST00000400371.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009270.hg.1 | ENST00000472805.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009270.hg.1 | ENST00000472805.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009270.hg.1 | ENST00000472805.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009270.hg.1 | ENST00000472805.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009270.hg.1 | NR_045983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009270.hg.1 | NR_045983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009270.hg.1 | NR_045983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009270.hg.1 | NR_045983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009270.hg.1 | NR_045983.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009270.hg.1 | NR_045983.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 221089 | - | - |
