Assay Details
| Gene Symbol: KIF16B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199865.1 | NP_001186794.1 | 20 - 21 | 3450 | 105 | |
| NM_024704.4 | NP_078980.3 | 21 - 22 | 3603 | 105 | ||
| XM_005260753.3 | XP_005260810.1 | 22 - 23 | 3613 | 105 | ||
| XM_005260754.3 | XP_005260811.1 | 22 - 23 | 3613 | 105 | ||
| XM_005260755.3 | XP_005260812.1 | 21 - 22 | 3506 | 105 | ||
| GenBank mRNA | AK026698.1 | - | 2 - 3 | 192 | 105 | |
| AY044654.1 | - | 1 - 2 | 105 | 105 | ||
| AY166853.1 | - | 21 - 22 | 3602 | 105 | ||
| BC034984.1 | - | 3 - 4 | 279 | 105 | ||
| BC050577.1 | - | 20 - 21 | 3450 | 105 | ||
| BX647572.1 | - | 21 - 22 | 3522 | 105 | ||
| BX648426.1 | - | 7 - 8 | 1308 | 105 | ||
More Information
Set Membership: |
|

KIF16B
Genomic Map