Assay Details
| Gene Symbol: CSN1S2AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003720.1 | 8 - 9 | 284 | 127 | ||
| GenBank mRNA | AY154892.1 | - | 8 - 9 | 284 | 127 | |
| BC128170.1 | - | 5 - 6 | 158 | 127 | ||
| BC128173.1 | - | 5 - 6 | 158 | 127 | ||
| BC141914.1 | - | 5 - 6 | 158 | 127 | ||
| BC150502.1 | - | 5 - 6 | 158 | 127 | ||
More Information
Set Membership: |
|

CSN1S2AP
Genomic Map