Assay Details
| Gene Symbol: LVRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173800.4 | NP_776161.3 | 14 - 15 | 2406 | 129 | |
| GenBank mRNA | AK075131.1 | - | 9 - 10 | 941 | 129 | |
| AK291655.1 | - | 14 - 15 | 2420 | 129 | ||
| AY560010.1 | - | 14 - 15 | 2294 | 129 | ||
| BC036440.1 | - | 9 - 10 | 1285 | 129 | ||
| BC060869.1 | - | 15 - 16 | 2333 | 30346049 | 129 | |
| BC070028.1 | - | 9 - 10 | 1403 | 129 | ||
| BC094716.1 | - | 9 - 10 | 1403 | 129 | ||
| BC109022.1 | - | 14 - 15 | 2270 | 40008501 | 129 | |
| BC109023.1 | - | 14 - 15 | 2270 | 40008502 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|

LVRN
Genomic Map