Assay Details
| Gene Symbol: OOSP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173801.4 | NP_776162.2 | 1 - 2 | 251 | 60 | |
| GenBank mRNA | AK093517.1 | - | 1 - 2 | 117 | 60 | |
| BC036256.1 | - | 1 - 2 | 117 | 5270384 | 60 | |
| BC048121.1 | - | 1 - 2 | 251 | 5266642 | 60 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000503.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11005965.hg.1 | ENST00000278855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005965.hg.1 | ENST00000532905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005965.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005965.hg.1 | TC11000503.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005962.hg.1 | ENST00000278855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005962.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005962.hg.1 | TC11000503.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | ENST00000527395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | ENST00000527395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | ENST00000527395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | ENST00000527395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | ENST00000532905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | ENST00000532905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | ENST00000532905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | ENST00000532905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007762.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005963.hg.1 | ENST00000278855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005963.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005963.hg.1 | TC11000503.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005964.hg.1 | ENST00000278855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005964.hg.1 | ENST00000532905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005964.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005964.hg.1 | TC11000503.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100154638.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154638.hg.1 | ENST00000527395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154638.hg.1 | ENST00000532905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154638.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154638.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154638.hg.1 | ENST00000527395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154638.hg.1 | ENST00000532905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154638.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154638.hg.1 | TC1100007762.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100154636.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154636.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154636.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154636.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154636.hg.1 | TC1100007762.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100154639.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154639.hg.1 | ENST00000527395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154639.hg.1 | ENST00000532905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154639.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154639.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154639.hg.1 | ENST00000527395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154639.hg.1 | ENST00000532905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154639.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154639.hg.1 | TC1100007762.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100154635.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154635.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154635.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154635.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154635.hg.1 | TC1100007762.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100154637.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154637.hg.1 | ENST00000527395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154637.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154637.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154637.hg.1 | ENST00000527395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154637.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154637.hg.1 | TC1100007762.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | ENST00000278855.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | ENST00000527395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | ENST00000527395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | ENST00000527395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | ENST00000527395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | ENST00000532905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | ENST00000532905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | ENST00000532905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | ENST00000532905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | NM_173801.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007762.hg.1 | NM_173801.4 | Refseq |

OOSP2
Genomic Map