Assay Details
| Gene Symbol: ANXA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156.4 | NP_001147.1 | 5 - 6 | 508 | 68 | |
| NM_001320879.1 | NP_001307808.1 | 4 - 5 | 453 | 68 | ||
| XM_017016163.1 | XP_016871652.1 | 5 - 6 | 544 | 68 | ||
| GenBank mRNA | AB062429.1 | - | 5 - 6 | 466 | 68 | |
| AK222552.1 | - | 5 - 6 | 476 | 68 | ||
| AK300086.1 | - | 3 - 4 | 275 | 68 | ||
| AK301681.1 | - | 5 - 6 | 262 | 68 | ||
| AK307772.1 | - | 4 - 5 | 327 | 68 | ||
| AK309141.1 | - | 5 - 6 | 270 | 68 | ||
| AK313024.1 | - | 5 - 6 | 451 | 68 | ||
| BC002632.2 | - | 5 - 6 | 468 | 3610541 | 68 | |
| BT007187.1 | - | 4 - 5 | 433 | 68 | ||
| BX361177.2 | - | 4 - 5 | 419 | 68 | ||
| CR407686.1 | - | 4 - 5 | 433 | 68 | ||
| J04543.1 | - | 5 - 6 | 493 | 68 | ||
More Information
Set Membership: |
|

ANXA7
Genomic Map