| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12005350.hg.1 |
ENST00000331817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12005350.hg.1 |
ENST00000422319.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12005350.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12005350.hg.1 |
TC12000418.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000546856.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000546856.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000546856.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000546856.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000546856.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000546856.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000546856.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000547613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000547613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000547613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000547613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000547613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000547613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000547613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000548088.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000548088.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000548088.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000548088.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000548088.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000548657.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000548657.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000548657.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000548657.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000549127.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000549127.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000549127.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000549127.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000549638.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000549638.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000549638.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000549638.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000549638.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000549638.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000550153.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000550153.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000550153.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000550153.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000550153.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000550153.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000551130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000551130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000551130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000551130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000551130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000552183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000552183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000552183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000552183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000552322.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000552322.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000552322.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000552322.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000552322.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000552322.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000552322.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000552322.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000552322.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
ENST00000552322.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200047.hg.1 |
ENST00000546856.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200047.hg.1 |
ENST00000551130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200047.hg.1 |
ENST00000552183.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200047.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200047.hg.1 |
ENST00000546856.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200047.hg.1 |
ENST00000551130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200047.hg.1 |
ENST00000552183.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200047.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200047.hg.1 |
TC1200012634.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000546856.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000546856.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000546856.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000546856.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000546856.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000546856.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000546856.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000547613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000547613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000547613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000547613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000547613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000547613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000547613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000548088.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000548088.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000548088.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000548088.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000548088.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000548657.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000548657.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000548657.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000548657.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000549127.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000549127.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000549127.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000549127.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000549638.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000549638.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000549638.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000549638.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000549638.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000549638.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000550153.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000550153.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000550153.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000550153.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000550153.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000550153.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000551130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000551130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000551130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000551130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000551130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000552183.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000552183.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000552183.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000552183.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000552322.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000552322.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000552322.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000552322.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000552322.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000552322.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000552322.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000552322.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000552322.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
ENST00000552322.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012634.hg.1 |
NM_005556.3 |
Refseq |