Assay Details
| Gene Symbol: KIAA0125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026800.2 | 6 | 9278 | 100 | ||
| GenBank mRNA | AW451894.1 | - | 1 | 100 | ||
| BC152473.1 | - | 1 | 7551 | 100 | ||
| D50915.1 | - | 1 | 7551 | 100 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09536392 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09536384 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09536404 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09536397 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16743841 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16743859 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09536399 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30003498 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09536382 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16743910 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09536396 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09536398 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16743908 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16743884 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09536395 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09536385 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30003415 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30003435 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16743882 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09536394 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09536406 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09536390 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30003478 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30003490 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09536408 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 76120 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 8868 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 131557 | AK131492.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 131557 | XM_379262.3 | refseq_mrna_human_XM |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 131557 | XM_940851.1 | refseq_mrna_human_XM |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 50308 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 5930 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 101545 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 672689 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 52787 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 262533 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 556682 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 145642 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 822862 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 187029 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 52686 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 262545 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 160023 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 170447 | AB040934.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 170447 | XM_290799.7 | refseq_mrna_human_XM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 170447 | XM_936596.2 | refseq_mrna_human_XM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 300373 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 300810 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 275170 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 397830 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 76337 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P95723 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P140190 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P95723 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 29314 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 23420 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 15906 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 16694 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P12134750 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_14_P136198 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P140190 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P95723 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P95723 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P140190 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | ERCC-00044_175 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743745 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743754 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536373 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536377 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30003347 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30003395 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536375 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536378 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743784 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536372 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536376 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536379 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536380 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30003363 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536381 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743731 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743792 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743771 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536383 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743783 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30003415 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743734 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30003315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536382 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743748 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743736 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30003347 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536380 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536375 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30003315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743745 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536379 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536373 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743754 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743771 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536382 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536377 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536372 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743734 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30003363 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30003395 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743736 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743792 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536383 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743784 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743748 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536381 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743783 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536376 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09536378 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30003415 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16743731 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P95723 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P136198 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P135714 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P95723 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536392 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536429 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16743739 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536400 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16743747 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536384 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536445 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536412 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16743733 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536380 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536382 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536452 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536440 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536376 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16743929 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536379 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16743806 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16743876 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16743744 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536395 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16743754 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536406 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536426 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16743749 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536402 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536425 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536437 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536375 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536417 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536455 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536409 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536457 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536449 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536435 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16743772 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536421 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536433 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536398 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536424 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536373 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536413 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536404 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536389 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536447 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536442 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536415 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09536439 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P136198 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P12134750 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P135714 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7977444 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7977446 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7977443 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7977445 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7977440 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 206478_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 206478_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 206478_at | - | - |
| Human Genome U133A 2.0 Array | 206478_PM_at | - | - |
| Human Genome U133A 2.0 Array | 206478_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw596479 | - | - |
| Human Genome U95 Set | 33528_at | - | - |
| Human Genome U95 Set | 33528_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 206478_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 206478_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 206478_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 206478_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 206478_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 206478_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 206478_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 206478_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 206478_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 206478_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 206478_at10 | - | - |
| Human Genome U95 Set | 66761_at | - | - |
| Human Genome U95 Set | 66761_PM_at | - | - |
| Human Genome U95 Set | 66762_PM_g_at | - | - |
| Human Genome U95 Set | 66762_g_at | - | - |
| Human Genome U133 Set | 206478_at | - | - |
| Human Genome U133 Set | 206478_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051875 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051875 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 649805 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1707491 | NR_026800.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1707491 | AW451894.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1707491 | BC152473.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1707491 | D50915.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1679196 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1679196 | - | - |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1679196 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1707491 | NR_026800.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1707491 | AW451894.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1707491 | BC152473.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1707491 | D50915.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1707491 | NR_026800.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1707491 | AW451894.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1707491 | BC152473.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1707491 | D50915.1 | human_transcriptome |

KIAA0125
Genomic Map