Assay Details
| Gene Symbol: RBM12B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027259.1 | 1 | 266 | 60 | ||
| GenBank mRNA | AF116672.1 | - | 1 | 256 | 60 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000568.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P134729 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P134729 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 27554 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 17475 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P134729 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P134729 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305631 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588097 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588132 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588125 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588120 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588099 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588084 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305657 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305622 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588098 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588123 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588111 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588127 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305643 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588109 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588095 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305671 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305644 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305691 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588094 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305669 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588082 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305692 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588068 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305642 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588080 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588110 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588071 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588113 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588129 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588101 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588076 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588064 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305639 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588078 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588065 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588121 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588126 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305661 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588088 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305679 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588070 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305667 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588073 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305677 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588106 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588067 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305684 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588119 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588105 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305687 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588125 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588099 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588073 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588109 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588080 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305622 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588120 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588064 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588119 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588098 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305692 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305657 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588101 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588082 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305679 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588088 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588095 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588084 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588068 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588132 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305644 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588094 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588127 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588110 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588111 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305643 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588067 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305639 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588065 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305642 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588105 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305691 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588123 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305631 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588129 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588078 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588076 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588070 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588071 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305671 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588121 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588126 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305661 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588106 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305684 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305677 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305669 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588113 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26588097 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305667 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06305687 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P134729 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305650 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305638 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305621 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305664 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305690 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305670 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305673 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305634 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305678 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305685 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305639 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305688 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16116918 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16116927 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305623 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305633 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305680 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305655 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305642 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305657 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305669 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305659 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305667 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305718 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305652 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305681 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305676 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305692 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305627 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305618 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305645 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06305629 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 220222_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007741.hg.1 | TC08000568.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007741.hg.1 | ENST00000391680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08007741.hg.1 | NR_027259.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008238.hg.1 | ENST00000623283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008238.hg.1 | NR_027259.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008238.hg.1 | NR_027259.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156564.hg.1 | ENST00000623283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156564.hg.1 | NR_027259.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156564.hg.1 | ENST00000623283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800156564.hg.1 | NR_027259.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800156564.hg.1 | TC0800008238.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800008238.hg.1 | ENST00000623283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008238.hg.1 | NR_027259.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008238.hg.1 | NR_027259.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 220222_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 220222_at | - | - |
| Human Genome U133A 2.0 Array | 220222_at | - | - |
| Human Genome U133A 2.0 Array | 220222_PM_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw599928 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220222_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220222_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220222_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220222_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220222_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220222_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220222_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220222_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220222_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220222_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220222_at9 | - | - |
| Human Genome U95 Set | 85918_PM_at | - | - |
| Human Genome U95 Set | 85918_at | - | - |
| Human Genome U133 Set | 220222_PM_at | - | - |
| Human Genome U133 Set | 220222_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036760 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036760 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 162661 | - | - |

RBM12B-AS1
Genomic Map