Assay Details
| Gene Symbol: GABBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021904.3 | NP_068704.2 | 3 - 4 | 573 | 70 | |
| XM_006715047.3 | XP_006715110.1 | 3 - 4 | 374 | 70 | ||
| GenBank mRNA | AJ012187.1 | - | 2 - 3 | 287 | 70 | |
| AL539611.3 | - | 3 - 4 | 573 | 70 | ||
More Information
Set Membership: |
|

GABBR1
Genomic Map