Assay Details
| Gene Symbol: PSMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002812.4 | NP_002803.2 | 3 - 4 | 584 | 70 | |
| GenBank mRNA | AK301771.1 | - | 3 - 4 | 595 | 70 | |
| AK308809.1 | - | 3 - 4 | 673 | 70 | ||
| BC001164.1 | - | 3 - 4 | 335 | 3528096 | 70 | |
| BC065006.1 | - | 3 - 4 | 323 | 6055235 | 70 | |
| BI560993.1 | - | 3 - 4 | 568 | 70 | ||
| BQ684248.1 | - | 3 - 4 | 347 | 70 | ||
| BT019688.1 | - | 3 - 4 | 253 | 70 | ||
| D38047.1 | - | 3 - 4 | 323 | 70 | ||
| EU668354.1 | - | 3 - 4 | 584 | 70 | ||
More Information
Set Membership: |
|

PSMD8
Genomic Map