Assay Details
Gene Symbol: HIST3H2BB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175055.2 | NP_778225.1 | 1 | 43 | 86 | |
GenBank mRNA | AK091220.1 | - | 1 | 43 | 86 | |
BC100855.1 | - | 1 | 42 | 40002412 | 86 | |
BC100856.1 | - | 1 | 42 | 40002414 | 86 | |
BC100857.1 | - | 1 | 42 | 40002416 | 86 | |
BC100858.1 | - | 1 | 42 | 40002418 | 86 | |
BC137467.1 | - | 1 | 42 | 9021467 | 86 | |
BC137468.1 | - | 1 | 42 | 9021468 | 86 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001893.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK091220.1,BC100855.1,BC100856.1,BC100857.1,BC100858.1,BC137467.1,BC137468.1,NM_175055.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846054 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15189089 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846049 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916290 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916324 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846045 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916327 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846051 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916302 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916297 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846052 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846061 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916291 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846039 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846062 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916325 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846053 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846050 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916308 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916307 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846063 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916303 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846029 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916298 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846036 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846027 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846046 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846022 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916313 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846024 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846060 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916294 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916312 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846059 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3229335 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P332992 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 42188 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 32563 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P128402 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P128402 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 22027 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 34694 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_14_P128402 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3229335 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3229335 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846044 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846046 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846036 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916313 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916327 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846049 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916325 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846022 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916303 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15189089 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846050 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846039 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15189099 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916324 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916298 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916291 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846052 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916302 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846059 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916308 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846029 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846032 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846054 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846028 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846053 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916312 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846045 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916294 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916290 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846061 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916290 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846039 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846044 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916291 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846045 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846054 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846061 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846029 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15189089 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846050 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846049 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916312 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15189099 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846059 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846046 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846028 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916313 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846032 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916324 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916303 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846052 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916302 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916294 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916327 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916298 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846053 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846036 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00846022 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916325 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20916308 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P332992 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P524452 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P128402 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00846042 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00846031 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00846028 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00846048 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00846055 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00846026 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00846057 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00846049 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00846059 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00846046 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00846040 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00846044 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00846045 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00846034 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00846052 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00846053 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00846036 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00846029 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P00846026 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15189098 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15189092 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P00846053 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P00846023 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15189100 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P00846050 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15189099 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P00846049 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15189091 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P00846024 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P00846025 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P00846052 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P00846051 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P128402 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P128402 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P332992 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P443407 | - | - |
Custom Human Agilent Array 11k, Chromosome 6, Expression | 3838 | - | - |
Custom Human Agilent Array 11k, Chromosome 6, Expression | 8897 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P771334 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P443407 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P915784 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846028 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846040 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846063 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846026 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846055 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15189089 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846042 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15189100 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15189091 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846036 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846031 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846049 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846059 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846048 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846022 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846061 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846045 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846044 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846024 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846034 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846058 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15189086 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846029 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846051 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846046 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15189098 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_175055_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7910371 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7910370 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7910369 | - | - |
Affymetrix Human Genome U219 Array | 11732990_x_at | - | - |
Affymetrix Human Genome U219 Array | 11732989_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11732990_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11732989_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR01029514.hg.1 | TC01001893.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01029514.hg.1 | ENST00000369160.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01029514.hg.1 | NM_175055.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011871.hg.1 | ENST00000620438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011871.hg.1 | ENST00000620438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011871.hg.1 | ENST00000620438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011871.hg.1 | NM_175055.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011871.hg.1 | NM_175055.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100187691.hg.1 | ENST00000620438.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100187691.hg.1 | NM_175055.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100187691.hg.1 | ENST00000620438.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100187691.hg.1 | NM_175055.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100187691.hg.1 | TC0100011871.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100187690.hg.1 | ENST00000620438.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100187690.hg.1 | NM_175055.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100187690.hg.1 | ENST00000620438.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100187690.hg.1 | NM_175055.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100187690.hg.1 | TC0100011871.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100011871.hg.1 | ENST00000620438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011871.hg.1 | ENST00000620438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011871.hg.1 | ENST00000620438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011871.hg.1 | NM_175055.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011871.hg.1 | NM_175055.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041380 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041380 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 172904 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1764986 | NM_175055.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1764986 | AK091220.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1764986 | NM_175055.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1764986 | AK091220.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1764986 | NM_175055.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1764986 | AK091220.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1764986 | NM_175055.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1764986 | AK091220.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1764986 | NM_175055.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1764986 | AK091220.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_28872748-S | NM_175055.2 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_28872748-S | AK091220.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_13915-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0006370097 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0006370097 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1764986 | NM_175055.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1764986 | AK091220.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1764986 | NM_175055.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1764986 | AK091220.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 1990376 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_13915 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1764986 | NM_175055.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1764986 | AK091220.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1764986 | NM_175055.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1764986 | AK091220.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1764986 | NM_175055.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1764986 | AK091220.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1764986 | NM_175055.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1764986 | AK091220.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1764986 | NM_175055.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1764986 | AK091220.1 | human_transcriptome |
