Assay Details
| Gene Symbol: ARL14EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152316.2 | NP_689529.1 | 1 - 2 | 111 | 154 | |
| XM_005252792.3 | XP_005252849.1 | 1 - 2 | 136 | 154 | ||
| GenBank mRNA | AK096287.1 | - | 1 - 2 | 108 | 154 | |
| BC047775.1 | - | 1 - 2 | 97 | 6062049 | 156 | |
| BP341330.1 | - | 1 - 2 | 111 | 156 | ||
| BX647617.1 | - | 1 - 2 | 101 | 156 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000295.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK096287.1,BC047775.1,BX647617.1,NM_152316.2,XM_005252792.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11003806.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003806.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003806.hg.1 | TC11000295.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003805.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003805.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003805.hg.1 | TC11000295.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003810.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003810.hg.1 | ENST00000533457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003810.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003810.hg.1 | TC11000295.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | ENST00000530909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | ENST00000530909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | ENST00000530909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | ENST00000530909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | ENST00000532047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | ENST00000532047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | ENST00000532047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | ENST00000532047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | ENST00000533457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | ENST00000533457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007168.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007250.hg.1 | ENST00000412776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003808.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003808.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003808.hg.1 | TC11000295.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003811.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003811.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003811.hg.1 | TC11000295.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003812.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003812.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003812.hg.1 | TC11000295.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100150422.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150422.hg.1 | ENST00000530909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150422.hg.1 | ENST00000533457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150422.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150422.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150422.hg.1 | ENST00000530909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150422.hg.1 | ENST00000533457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150422.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150422.hg.1 | TC1100007168.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | ENST00000530909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | ENST00000530909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | ENST00000530909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | ENST00000530909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | ENST00000532047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | ENST00000532047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | ENST00000532047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | ENST00000532047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | ENST00000533457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | ENST00000533457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007168.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150416.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150416.hg.1 | ENST00000532047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150416.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150416.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150416.hg.1 | ENST00000532047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150416.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150416.hg.1 | TC1100007168.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100150419.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150419.hg.1 | ENST00000530909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150419.hg.1 | ENST00000532047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150419.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150419.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150419.hg.1 | ENST00000530909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150419.hg.1 | ENST00000532047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150419.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150419.hg.1 | TC1100007168.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100150424.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150424.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150424.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150424.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150424.hg.1 | TC1100007168.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100150425.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150425.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150425.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150425.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150425.hg.1 | TC1100007168.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100150418.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150418.hg.1 | ENST00000530909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150418.hg.1 | ENST00000532047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150418.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150418.hg.1 | ENST00000282032.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150418.hg.1 | ENST00000530909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150418.hg.1 | ENST00000532047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150418.hg.1 | NM_152316.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150418.hg.1 | TC1100007168.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200007250.hg.1 | ENST00000412776.1 | Ensembl |

ARL14EP
Genomic Map