Assay Details
| Gene Symbol: PAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006451.4 | NP_006442.2 | 7 - 8 | 1323 | 137 | |
| NM_182789.3 | NP_877590.1 | 7 - 8 | 1086 | 137 | ||
| NM_183323.2 | NP_899152.1 | 7 - 8 | 891 | 137 | ||
| XM_005248230.3 | XP_005248287.1 | 7 - 8 | 1045 | 137 | ||
| XM_017008956.1 | XP_016864445.1 | 7 - 8 | 1226 | 137 | ||
| GenBank mRNA | AF013758.2 | - | 7 - 8 | 1263 | 137 | |
| AK308270.1 | - | 7 - 8 | 1086 | 137 | ||
| AL136920.1 | - | 7 - 8 | 1270 | 137 | ||
| BC005295.1 | - | 7 - 8 | 1026 | 4052678 | 137 | |
| BC015937.2 | - | 7 - 8 | 1309 | 3922262 | 137 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001320.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000306846.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000306846.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000306846.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000306846.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000306846.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000306846.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000306846.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000306846.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000306846.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000306846.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000306846.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000306846.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000306846.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000306846.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000306846.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000306846.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000306846.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000306846.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000306846.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000306846.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000338972.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000338972.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000338972.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000338972.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000338972.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000338972.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000338972.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000338972.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000338972.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000338972.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000338972.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000338972.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000338972.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000338972.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000338972.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000338972.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000338972.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000338972.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000436644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000436644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000436644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000436644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000436644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000436644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000436644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000436644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000436644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000436644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000436644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000436644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000436644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000436644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000436644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000436644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000436644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000436644.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000504075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000504075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000504075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000504075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000504639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000504639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000504639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000504639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000504639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000504639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000504639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000504639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000504639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000504639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000504639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000504639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000504639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000504639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000504639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000504639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000504639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000508537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000508537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000508537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000508537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000508537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000508537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000508537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000508537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000511321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000511321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000511321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000511321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000511321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000511321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000511321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000511321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000511321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514816.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514816.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514816.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514816.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000514816.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000515338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000515338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000515338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000515338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000515338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | ENST00000515338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_006451.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_006451.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_006451.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_006451.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_006451.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_006451.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_006451.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_006451.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_006451.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_006451.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_006451.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_006451.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_006451.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_006451.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_006451.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_006451.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_006451.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_006451.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_006451.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_006451.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_182789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_182789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_182789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_182789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_182789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_182789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_182789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_182789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_182789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_182789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_182789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_182789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_182789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_182789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_182789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_182789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_182789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_182789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_182789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_183323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_183323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_183323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_183323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_183323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_183323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_183323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_183323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_183323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_183323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_183323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_183323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_183323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_183323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_183323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_183323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_183323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010643.hg.1 | NM_183323.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 188840 | - | - |

PAIP1
Genomic Map