Assay Details
| Gene Symbol: GGTLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178311.2 | NP_842563.1 | 1 - 2 | 173 | 95 | |
| XM_005260863.4 | XP_005260920.1 | 1 - 2 | 528 | 95 | ||
| GenBank mRNA | BC040904.1 | - | 1 - 2 | 173 | 5744021 | 95 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000721.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC040904.1,NM_178311.2,XM_005260863.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000278765.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000278765.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000278765.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000278765.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000278765.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000278765.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000278765.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000278765.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000286890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000286890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000286890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000286890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000286890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000286890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000286890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000286890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000335694.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000335694.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000335694.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000335694.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000335694.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000335694.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000335694.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | ENST00000335694.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | NM_178311.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | NM_178311.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | NM_178311.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | NM_178311.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | NM_178311.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | NM_178311.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | NM_178311.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | NM_178311.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | NM_178312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | NM_178312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | NM_178312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | NM_178312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | NM_178312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | NM_178312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | NM_178312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008690.hg.1 | NM_178312.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000417145.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000417145.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000417145.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000417145.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000417145.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000417145.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000417145.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000417145.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000417145.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000417145.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000448514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000448514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000448514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000448514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000448514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000448514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000480559.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000480559.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000480559.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000480559.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000480559.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000480559.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000613850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000613850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000613850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000613850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000613850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000613850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000613850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000613850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000613850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000618722.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000618722.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000618722.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000618722.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000618722.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000618722.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000618722.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000618722.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000618722.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | ENST00000618722.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | NM_199127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | NM_199127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | NM_199127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | NM_199127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | NM_199127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | NM_199127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | NM_199127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006793.hg.1 | NM_199127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20009791.hg.1 | ENST00000335694.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20009791.hg.1 | NM_178311.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20009791.hg.1 | TC20000721.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000160827.hg.1 | ENST00000335694.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000160827.hg.1 | NM_178311.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000160827.hg.1 | TC2000008690.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000160827.hg.1 | ENST00000335694.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000160827.hg.1 | NM_178311.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000278765.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000278765.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000278765.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000278765.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000278765.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000278765.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000278765.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000278765.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000286890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000286890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000286890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000286890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000286890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000286890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000286890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000286890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000335694.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000335694.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000335694.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000335694.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000335694.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000335694.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000335694.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | ENST00000335694.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | NM_178311.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | NM_178311.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | NM_178311.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | NM_178311.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | NM_178311.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | NM_178311.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | NM_178311.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | NM_178311.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | NM_178312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | NM_178312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | NM_178312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | NM_178312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | NM_178312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | NM_178312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | NM_178312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008690.hg.1 | NM_178312.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000417145.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000417145.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000417145.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000417145.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000417145.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000417145.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000417145.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000417145.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000417145.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000417145.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000448514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000448514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000448514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000448514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000448514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000448514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000480559.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000480559.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000480559.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000480559.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000480559.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000480559.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000613850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000613850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000613850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000613850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000613850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000613850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000613850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000613850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000613850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000618722.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000618722.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000618722.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000618722.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000618722.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000618722.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000618722.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000618722.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000618722.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | ENST00000618722.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | NM_001282879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | NM_199127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | NM_199127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | NM_199127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | NM_199127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | NM_199127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | NM_199127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | NM_199127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006793.hg.1 | NM_199127.2 | Refseq |

GGTLC1
Genomic Map