Assay Details
| Gene Symbol: MKI67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145966.1 | NP_001139438.1 | 10 - 11 | 1374 | 137 | |
| NM_002417.4 | NP_002408.3 | 11 - 12 | 2454 | 137 | ||
| XM_011539818.2 | XP_011538120.1 | 8 - 9 | 2185 | 137 | ||
| GenBank mRNA | AJ567756.1 | - | 7 - 8 | 984 | 137 | |
| X65550.1 | - | 11 - 12 | 2455 | 137 | ||
| X65551.1 | - | 10 - 11 | 1375 | 137 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001756.hg.1
Set Membership: |
|

MKI67
Genomic Map