Assay Details
| Gene Symbol: CD53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000560.3 | NP_000551.1 | 2 - 3 | 228 | 95 | |
| NM_001040033.1 | NP_001035122.1 | 3 - 4 | 233 | 95 | ||
| NM_001320638.1 | NP_001307567.1 | 2 - 3 | 228 | 95 | ||
| GenBank mRNA | AK298723.1 | - | 2 - 3 | 175 | 95 | |
| AK313582.1 | - | 2 - 3 | 181 | 95 | ||
| BC035456.1 | - | 3 - 4 | 215 | 95 | ||
| BC040693.1 | - | 2 - 3 | 158 | 5498854 | 95 | |
| BG536920.1 | - | 2 - 3 | 229 | 95 | ||
| BT019643.1 | - | 1 - 2 | 62 | 95 | ||
| CA453414.1 | - | 235 | 95 | |||
| M37033.1 | - | 2 - 3 | 155 | 95 | ||
| M60871.1 | - | 2 - 3 | 135 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000968.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400008109.hg.1 | ENST00000503756.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008109.hg.1 | ENST00000503756.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015414.hg.1 | TC01000968.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015414.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015424.hg.1 | TC01000968.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015424.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015424.hg.1 | ENST00000429072.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015424.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015424.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015423.hg.1 | TC01000968.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015423.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015423.hg.1 | ENST00000429072.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015423.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015423.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000464329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000464329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000464329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000464329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015422.hg.1 | TC01000968.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015422.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015422.hg.1 | ENST00000429072.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015422.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015422.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167535.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167535.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167535.hg.1 | TC0100009394.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000464329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000464329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000464329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000464329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167543.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167543.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167543.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167543.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167543.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167543.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167543.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167543.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167543.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167543.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167543.hg.1 | TC0100009394.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100167542.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167542.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167542.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167542.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167542.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167542.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167542.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167542.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167542.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167542.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167542.hg.1 | TC0100009394.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400008109.hg.1 | ENST00000503756.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008109.hg.1 | ENST00000503756.2 | Ensembl |

CD53
Genomic Map