Assay Details
| Gene Symbol: CELSR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001407.2 | NP_001398.2 | 5 - 6 | 5116 | 87 | |
| GenBank mRNA | AF231023.1 | - | 5 - 6 | 5116 | 87 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03003382.hg.1
Set Membership: |
|

CELSR3
Genomic Map