Assay Details
| Gene Symbol: GCKR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001486.3 | NP_001477.2 | 8 - 9 | 711 | 68 | |
| XM_011532763.1 | XP_011531065.1 | 8 - 9 | 711 | 68 | ||
| XM_017003796.1 | XP_016859285.1 | 3 - 4 | 188 | 68 | ||
| XM_017003797.1 | XP_016859286.1 | 2 - 3 | 138 | 68 | ||
| GenBank mRNA | AK298448.1 | - | 7 - 8 | 635 | 68 | |
| AK303784.1 | - | 7 - 8 | 635 | 68 | ||
| AK309599.1 | - | 8 - 9 | 667 | 68 | ||
| BC130481.1 | - | 8 - 9 | 691 | 40146512 | 68 | |
| BC130483.1 | - | 8 - 9 | 691 | 40146514 | 68 | |
| Z48475.1 | - | 8 - 9 | 708 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000176.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein import into nucleus, translocation
carbohydrate metabolic process
response to fructose
regulation of glucose transport
negative regulation of glucokinase activity
urate metabolic process
triglyceride homeostasis
protein binding
carbohydrate binding
fructose-6-phosphate binding
protein import into nucleus, translocation
carbohydrate metabolic process
response to fructose
regulation of glucose transport
negative regulation of glucokinase activity
urate metabolic process
triglyceride homeostasis
protein binding
carbohydrate binding
fructose-6-phosphate binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000411584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000411584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000411584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000411584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000411584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000411584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000411584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000411584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000417872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000417872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000417872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000417872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000417872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000417872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000417872.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000453813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000453813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000453813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000453813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000453813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000453813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000453813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000453813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000453813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000478147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000478147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000478147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | ENST00000478147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02002376.hg.1 | TC02000176.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02002376.hg.1 | ENST00000264717.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02002376.hg.1 | ENST00000424318.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02002376.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148552.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148552.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148552.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148552.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148552.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200148552.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200148552.hg.1 | TC0200007070.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000264717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000411584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000411584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000411584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000411584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000411584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000411584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000411584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000411584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000417872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000417872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000417872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000417872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000417872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000417872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000417872.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000453813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000453813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000453813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000453813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000453813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000453813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000453813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000453813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000453813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000472290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000478147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000478147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000478147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | ENST00000478147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007070.hg.1 | NM_001486.3 | Refseq |

GCKR
Genomic Map