Assay Details
| Gene Symbol: IRF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002163.2 | NP_002154.1 | 2 - 3 | 236 | 102 | |
| XM_017023199.1 | XP_016878688.1 | 1 - 2 | 171 | 102 | ||
| GenBank mRNA | AK313524.1 | - | 2 - 3 | 601 | 102 | |
| BC126247.1 | - | 2 - 3 | 231 | 8991963 | 102 | |
| BG107396.1 | - | 2 - 3 | 218 | 102 | ||
| BI835080.1 | - | 2 - 3 | 235 | 102 | ||
| M91196.1 | - | 2 - 3 | 226 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017023199.1 transcripts.
Set Membership: |
|

IRF8
Genomic Map