Assay Details
| Gene Symbol: VAV2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134398.1 | NP_001127870.1 | 24 - 25 | 2060 | 60 | |
| NM_003371.3 | NP_003362.2 | 22 - 23 | 2030 | 60 | ||
| XM_005272213.1 | XP_005272270.1 | 22 - 23 | 2080 | 60 | ||
| XM_017015108.1 | XP_016870597.1 | 25 - 26 | 2164 | 60 | ||
| XM_017015109.1 | XP_016870598.1 | 24 - 25 | 2149 | 60 | ||
| XM_017015110.1 | XP_016870599.1 | 23 - 24 | 2095 | 60 | ||
| XM_017015111.1 | XP_016870600.1 | 25 - 26 | 2164 | 60 | ||
| XM_017015112.1 | XP_016870601.1 | 23 - 24 | 2095 | 60 | ||
| GenBank mRNA | AK124271.1 | - | 3 - 4 | 60 | ||
| BC033187.1 | - | 8 - 9 | 575 | 60 | ||
| BC128448.1 | - | 11 - 12 | 914 | 60 | ||
| BC132965.1 | - | 22 - 23 | 2030 | 40146987 | 60 | |
| BC132967.1 | - | 22 - 23 | 2030 | 40146989 | 60 | |
| BX640754.1 | - | 22 - 23 | 1870 | 60 | ||
| S76992.1 | - | 24 - 25 | 2019 | 60 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001696.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK124271.1 transcripts.
Set Membership: |
|

VAV2
Genomic Map