Assay Details
| Gene Symbol: TRADD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003789.3 | NP_003780.1 | 1 - 2 | 70 | 86 | |
| GenBank mRNA | AK315654.1 | - | 1 - 2 | 463 | 86 | |
| BG717460.1 | - | 1 - 2 | 65 | 86 | ||
| DA353575.1 | - | 1 - 2 | 70 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001180.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK315654.1,NM_003789.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
signal transduction
I-kappaB kinase/NF-kappaB signaling
extrinsic apoptotic signaling pathway via death domain receptors
regulation of tumor necrosis factor-mediated signaling pathway
regulation of necrotic cell death
tumor necrosis factor-mediated signaling pathway
positive regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of NF-kappaB transcription factor activity
protein heterooligomerization
positive regulation of hair follicle development
death-inducing signaling complex assembly
extrinsic apoptotic signaling pathway
activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
signal transducer activity
tumor necrosis factor receptor binding
protein binding
kinase binding
protein complex binding
identical protein binding
binding, bridging
death domain binding
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
signal transduction
I-kappaB kinase/NF-kappaB signaling
extrinsic apoptotic signaling pathway via death domain receptors
regulation of tumor necrosis factor-mediated signaling pathway
regulation of necrotic cell death
tumor necrosis factor-mediated signaling pathway
positive regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of NF-kappaB transcription factor activity
protein heterooligomerization
positive regulation of hair follicle development
death-inducing signaling complex assembly
extrinsic apoptotic signaling pathway
activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
signal transducer activity
tumor necrosis factor receptor binding
protein binding
kinase binding
protein complex binding
identical protein binding
binding, bridging
death domain binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16016394.hg.1 | ENST00000345057.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016394.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016394.hg.1 | TC16001180.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016395.hg.1 | ENST00000345057.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016395.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016395.hg.1 | TC16001180.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000486556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000486556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000486556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000486556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000486556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000486556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000486556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000486556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000486556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000486556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000563348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000563348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000563348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000563348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000566104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000566104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000566104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000566247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000566247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000566247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | ENST00000566247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010616.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000486556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000486556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000486556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000486556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000486556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000486556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000486556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000486556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000486556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000486556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000563348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000563348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000563348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000563348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000566104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000566104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000566104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000566247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000566247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000566247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | ENST00000566247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010616.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181668.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181668.hg.1 | ENST00000563348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181668.hg.1 | ENST00000566104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181668.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181668.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181668.hg.1 | ENST00000563348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181668.hg.1 | ENST00000566104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181668.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181668.hg.1 | TC1600010616.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600181671.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181671.hg.1 | ENST00000566104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181671.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181671.hg.1 | ENST00000345057.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181671.hg.1 | ENST00000566104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181671.hg.1 | NM_003789.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181671.hg.1 | TC1600010616.hg.1 | Clariom D TC |

TRADD
Genomic Map