Assay Details
| Gene Symbol: EDF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281297.1 | NP_001268226.1 | 1 - 2 | 157 | 87 | |
| NM_001281298.1 | NP_001268227.1 | 1 - 2 | 157 | 87 | ||
| NM_001281299.1 | NP_001268228.1 | 1 - 2 | 157 | 87 | ||
| NM_003792.3 | NP_003783.1 | 1 - 2 | 157 | 87 | ||
| NM_153200.2 | NP_694880.1 | 1 - 2 | 157 | 87 | ||
| GenBank mRNA | AB002282.1 | - | 1 - 2 | 116 | 87 | |
| AB002283.2 | - | 1 - 2 | 116 | 87 | ||
| BC015500.1 | - | 1 - 2 | 107 | 3903539 | 87 | |
| BM800379.1 | - | 1 - 2 | 119 | 87 | ||
| BQ279031.1 | - | 1 - 2 | 106 | 87 | ||
| BT009863.1 | - | 1 - 2 | 79 | 87 | ||
| CR541914.1 | - | 1 - 2 | 79 | 87 | ||
| HY029111.1 | - | 1 - 2 | 157 | 87 | ||
| HY170122.1 | - | 1 - 2 | 134 | 87 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001744.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
regulation of lipid metabolic process
positive regulation of DNA binding
endothelial cell differentiation
positive regulation of transcription, DNA-templated
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
histone acetyltransferase activity
protein binding
calmodulin binding
methyltransferase activity
sequence-specific DNA binding
poly(A) RNA binding
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
regulation of lipid metabolic process
positive regulation of DNA binding
endothelial cell differentiation
positive regulation of transcription, DNA-templated
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
histone acetyltransferase activity
protein binding
calmodulin binding
methyltransferase activity
sequence-specific DNA binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000224073.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000224073.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000224073.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000224073.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000224073.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000224073.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000224073.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281298.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281298.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281298.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281298.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281298.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281298.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281298.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281299.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281299.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281299.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281299.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281299.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281299.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_001281299.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012011.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022444.hg.1 | TC09001744.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022444.hg.1 | ENST00000224073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022444.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022444.hg.1 | ENST00000371649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022444.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022444.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022443.hg.1 | TC09001744.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022443.hg.1 | ENST00000224073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022443.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022443.hg.1 | ENST00000371649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022443.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022443.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022445.hg.1 | TC09001744.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022445.hg.1 | ENST00000224073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022445.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022445.hg.1 | ENST00000371649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022445.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022445.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182457.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182457.hg.1 | NM_001281298.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182457.hg.1 | NM_001281299.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182457.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182457.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182457.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182457.hg.1 | NM_001281298.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182457.hg.1 | NM_001281299.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182457.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182457.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182457.hg.1 | TC0900012011.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900182453.hg.1 | ENST00000224073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182453.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182453.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182453.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182453.hg.1 | NM_001281298.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182453.hg.1 | NM_001281299.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182453.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182453.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182453.hg.1 | ENST00000224073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182453.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182453.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182453.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182453.hg.1 | NM_001281298.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182453.hg.1 | NM_001281299.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182453.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182453.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182453.hg.1 | TC0900012011.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000224073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000224073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000224073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000224073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000224073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000224073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000224073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281298.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281298.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281298.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281298.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281298.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281298.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281298.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281299.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281299.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281299.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281299.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281299.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281299.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_001281299.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012011.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182452.hg.1 | ENST00000224073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182452.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182452.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182452.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182452.hg.1 | NM_001281298.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182452.hg.1 | NM_001281299.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182452.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182452.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182452.hg.1 | ENST00000224073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182452.hg.1 | ENST00000371648.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182452.hg.1 | ENST00000371649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182452.hg.1 | NM_001281297.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182452.hg.1 | NM_001281298.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182452.hg.1 | NM_001281299.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182452.hg.1 | NM_003792.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182452.hg.1 | NM_153200.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182452.hg.1 | TC0900012011.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 141422 | - | - |

EDF1
Genomic Map