Assay Details
| Gene Symbol: ADAMTS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014243.2 | NP_055058.2 | 20 - 21 | 2964 | 57 | |
| XM_011532421.2 | XP_011530723.1 | 20 - 21 | 2996 | 57 | ||
| XM_011532422.2 | XP_011530724.1 | 20 - 21 | 3495 | 57 | ||
| GenBank mRNA | AB002364.1 | - | 20 - 21 | 2916 | 57 | |
| BC016451.1 | - | 3 - 4 | 404 | 57 | ||
| BC130287.1 | - | 20 - 21 | 2961 | 40146318 | 57 | |
| BC132735.1 | - | 20 - 21 | 2961 | 40146757 | 57 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001277.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
vascular endothelial growth factor production
protein processing
collagen fibril organization
collagen catabolic process
collagen biosynthetic process
extracellular fibril organization
positive regulation of vascular endothelial growth factor signaling pathway
endopeptidase activity
metalloendopeptidase activity
protein binding
heparin binding
zinc ion binding
vascular endothelial growth factor production
protein processing
collagen fibril organization
collagen catabolic process
collagen biosynthetic process
extracellular fibril organization
positive regulation of vascular endothelial growth factor signaling pathway
endopeptidase activity
metalloendopeptidase activity
protein binding
heparin binding
zinc ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000505193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000505193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000505193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000505193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000511274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000511274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000511274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000511274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000511274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000511274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000511274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017402.hg.1 | TC04001277.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017402.hg.1 | ENST00000286657.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017402.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000505193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000505193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000505193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000505193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000511274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000511274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000511274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000511274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000511274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000511274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000511274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010989.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400174661.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174661.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174661.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400174661.hg.1 | ENST00000286657.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174661.hg.1 | ENST00000622135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174661.hg.1 | NM_014243.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400174661.hg.1 | TC0400010989.hg.1 | Clariom D TC |

ADAMTS3
Genomic Map