Assay Details
| Gene Symbol: LIME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017806.3 | NP_060276.2 | 4 - 5 | 536 | 129 | |
| GenBank mRNA | AK000413.1 | - | 4 - 5 | 353 | 129 | |
| BC017016.1 | - | 4 - 5 | 301 | 3847069 | 129 | |
| BP368475.1 | - | 4 - 5 | 370 | 129 | ||
More Information
Set Membership: |
|

LIME1
Genomic Map