Assay Details
| Gene Symbol: CASP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032974.4 | NP_116756.2 | 7 - 8 | 1235 | 76 | |
| NM_032977.3 | NP_116759.2 | 7 - 8 | 1235 | 76 | ||
| XM_005246907.2 | XP_005246964.1 | 7 - 8 | 1018 | 76 | ||
| GenBank mRNA | AF111345.1 | - | 6 - 7 | 817 | 76 | |
| BC042844.1 | - | 7 - 8 | 971 | 5176698 | 76 | |
| KU177959.1 | - | 6 - 7 | 817 | 76 | ||
| U86214.1 | - | 7 - 8 | 1011 | 76 | ||
More Information
Set Membership: |
|

CASP10
Genomic Map