Assay Details
| Gene Symbol: TNFRSF25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003790.2 | NP_003781.1 | 6 - 7 | 686 | 90 | |
| NM_148967.1 | NP_683868.1 | 5 - 6 | 551 | 90 | ||
| GenBank mRNA | AY358309.1 | - | 6 - 7 | 700 | 90 | |
| BC117189.1 | - | 6 - 7 | 666 | 40125740 | 90 | |
| U72763.1 | - | 6 - 7 | 598 | 90 | ||
| U74611.1 | - | 6 - 7 | 686 | 90 | ||
| U75380.1 | - | 6 - 7 | 589 | 90 | ||
| U78029.1 | - | 6 - 7 | 598 | 90 | ||
| U83597.1 | - | 6 - 7 | 582 | 90 | ||
| U94501.1 | - | 6 - 7 | 598 | 90 | ||
| U94502.1 | - | 6 - 7 | 598 | 90 | ||
| U94509.1 | - | 5 - 6 | 463 | 90 | ||
More Information
Set Membership: |
|

TNFRSF25
Genomic Map