Assay Details
| Gene Symbol: FLJ31713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK056275.1 | - | 1 | 2550 | 62 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908085 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908087 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908114 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908105 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908125 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908116 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27252173 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908098 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908109 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27252208 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27252166 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16255473 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908103 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908115 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908113 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27252193 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27252197 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16255472 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908091 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908129 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908090 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908110 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908111 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908121 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908127 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908119 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908117 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908095 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908107 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908099 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27252206 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908086 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908096 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908084 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908097 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16255471 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908083 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908100 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908089 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27252188 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06908101 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16255476 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27252211 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27252191 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27252215 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 111476 | AK090672.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 111476 | BC100814.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 111476 | BC100815.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 111476 | BC100816.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 111476 | BC100817.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 111476 | NM_177402.3 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 10250 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 137831 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3318327 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P354326 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 31389 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P112258 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P112258 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 59515 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_14_P112258 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3318327 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P354326 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P112258 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P112258 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P112258 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P354326 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 31550 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 98860 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P112258 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P112258 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | XR_041883_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | XR_041882_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | XR_041881_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8163534 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8163533 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1553429_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1553429_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1553429_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553429_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553429_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553429_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553429_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553429_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553429_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553429_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553429_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553429_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553429_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553429_at8 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 218042 | - | - |

FLJ31713
Genomic Map