Assay Details
| Gene Symbol: GOLGA2P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120609.1 | 2 - 3 | 926 | 60 | ||
| GenBank mRNA | AK097852.1 | - | 3 - 4 | 423 | 60 | |
| AK125739.1 | - | 2 - 3 | 926 | 60 | ||
| AK309583.1 | - | 2 - 3 | 519 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000408070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000611134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | ENST00000611134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_106985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_106985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186876.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186876.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186876.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186876.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186876.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186876.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186876.hg.1 | TC1000012539.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000186877.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186877.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186877.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186877.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186877.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186877.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186877.hg.1 | TC1000012539.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000408070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000611134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | ENST00000611134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_106985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_106985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012539.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186867.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186867.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186867.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186867.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186867.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186867.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186867.hg.1 | TC1000012539.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000186872.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186872.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186872.hg.1 | TC1000012539.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000186869.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186869.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186869.hg.1 | ENST00000611134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186869.hg.1 | NR_106985.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186869.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186869.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186869.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186869.hg.1 | ENST00000611134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186869.hg.1 | NR_106985.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186869.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186869.hg.1 | TC1000012539.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000186881.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186881.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186881.hg.1 | TC1000012539.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000186866.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186866.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186866.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186866.hg.1 | ENST00000340929.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186866.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186866.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186866.hg.1 | TC1000012539.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000186870.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186870.hg.1 | ENST00000611134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186870.hg.1 | NR_106985.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186870.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186870.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186870.hg.1 | ENST00000611134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186870.hg.1 | NR_106985.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186870.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186870.hg.1 | TC1000012539.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000186874.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186874.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186874.hg.1 | ENST00000471055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186874.hg.1 | NR_120609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186874.hg.1 | TC1000012539.hg.1 | Clariom D TC |

GOLGA2P6
Genomic Map