Assay Details
| Gene Symbol: METTL25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319675.1 | NP_001306604.1 | 8 - 9 | 1644 | 131 | |
| NM_032230.2 | NP_115606.2 | 8 - 9 | 1552 | 131 | ||
| XM_011538827.2 | XP_011537129.1 | 7 - 8 | 1340 | 131 | ||
| XM_011538828.2 | XP_011537130.1 | 8 - 9 | 1552 | 131 | ||
| XM_011538829.2 | XP_011537131.1 | 8 - 9 | 1552 | 131 | ||
| XM_017020020.1 | XP_016875509.1 | 9 - 10 | 1615 | 131 | ||
| XM_017020021.1 | XP_016875510.1 | 8 - 9 | 2535 | 131 | ||
| GenBank mRNA | AK310231.1 | - | 8 - 9 | 1520 | 131 | |
| BC029120.1 | - | 8 - 9 | 1504 | 5189049 | 131 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000677.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12008910.hg.1 | ENST00000248306.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12008910.hg.1 | ENST00000547357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12008910.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12008910.hg.1 | TC12000677.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000547985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000547985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000547985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000548200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000548200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000548200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000548200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000548200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000548432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000548432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000548432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000548432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000548432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000548432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000548432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000548569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000548569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000548569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000550058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000550058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000550058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000550058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000550298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000550298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000550298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000550298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000550298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000550298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000551645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000551645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000551645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000551645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000551645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000551722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000551722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000551722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000551722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000551722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000552741.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000552741.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000552741.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000552741.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000552741.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | ENST00000552741.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000547985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000547985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000547985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000548200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000548200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000548200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000548200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000548200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000548432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000548432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000548432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000548432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000548432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000548432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000548432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000548569.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000548569.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000548569.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000550058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000550058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000550058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000550058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000550298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000550298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000550298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000550298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000550298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000550298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000551645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000551645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000551645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000551645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000551645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000551722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000551722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000551722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000551722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000551722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000552741.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000552741.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000552741.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000552741.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000552741.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | ENST00000552741.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008318.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200161235.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200161235.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200161235.hg.1 | ENST00000548432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200161235.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200161235.hg.1 | ENST00000248306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200161235.hg.1 | ENST00000547357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200161235.hg.1 | ENST00000548432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200161235.hg.1 | NM_032230.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200161235.hg.1 | TC1200008318.hg.1 | Clariom D TC |

METTL25
Genomic Map