Assay Details
| Gene Symbol: GPR152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206997.1 | NP_996880.1 | 1 | 356 | 115 | |
| GenBank mRNA | AY255600.1 | - | 1 | 72 | 115 | |
| AY569571.1 | - | 1 | 356 | 115 | ||
| BC119779.1 | - | 1 | 356 | 40109407 | 115 | |
| BC122869.1 | - | 1 | 356 | 40109404 | 115 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001984.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100011335.hg.1 | ENST00000312457.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011335.hg.1 | NM_206997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000602912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11023835.hg.1 | ENST00000312457.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11023835.hg.1 | NM_206997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11023835.hg.1 | TC11001984.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000602912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100187339.hg.1 | ENST00000312457.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100187339.hg.1 | NM_206997.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100187339.hg.1 | ENST00000312457.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100187339.hg.1 | NM_206997.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100187339.hg.1 | TC1100011335.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100011335.hg.1 | ENST00000312457.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011335.hg.1 | NM_206997.1 | Refseq |

GPR152
Genomic Map