Assay Details
| Gene Symbol: ZNF34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286770.1 | NP_001273699.1 | 4 - 5 | 326 | 93 | |
| XM_011517320.2 | XP_011515622.1 | 4 - 5 | 356 | 93 | ||
| XM_017013876.1 | XP_016869365.1 | 4 - 5 | 310 | 93 | ||
| GenBank mRNA | BC028136.1 | - | 2 - 3 | 548 | 5215496 | 93 |
| BP199961.1 | - | 4 - 5 | 326 | 93 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000534337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000534337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000534337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000534337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000534337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000534337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000534445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000534445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000534445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000534445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000534445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | ENST00000534445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012237.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184053.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184053.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184053.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184053.hg.1 | ENST00000534337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184053.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184053.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184053.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184053.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184053.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184053.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184053.hg.1 | ENST00000534337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184053.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184053.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184053.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184053.hg.1 | TC0800012237.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800184057.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184057.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184057.hg.1 | TC0800012237.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800184048.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184048.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184048.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184048.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184048.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184048.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184048.hg.1 | TC0800012237.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800184051.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184051.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184051.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184051.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184051.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184051.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184051.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184051.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184051.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184051.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184051.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184051.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184051.hg.1 | TC0800012237.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800184052.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184052.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184052.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184052.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184052.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184052.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184052.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184052.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184052.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184052.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184052.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184052.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184052.hg.1 | TC0800012237.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000343459.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000429371.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000527740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000534337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000534337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000534337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000534337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000534337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000534337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000534445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000534445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000534445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000534445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000534445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | ENST00000534445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_001286769.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_001286770.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_030580.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012237.hg.1 | NM_030580.4 | Refseq |

ZNF34
Genomic Map