Assay Details
Gene Symbol: LOC728175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040108.1 | 2 - 3 | 849 | 74 | ||
GenBank mRNA | AK097110.1 | - | 2 - 3 | 849 | 74 | |
BC020886.1 | - | 2 - 3 | 748 | 74 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001803.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC020886.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843609 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843608 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843587 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682582 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682586 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682606 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682559 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682556 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682624 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682573 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682563 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843562 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843571 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682543 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682620 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843666 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682547 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682627 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682622 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682545 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682594 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843637 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682597 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682619 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682638 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843588 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682535 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843617 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843655 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843656 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843590 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843560 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843631 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843565 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682600 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682640 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682575 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843642 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682541 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682572 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682528 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843645 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843598 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843582 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682636 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843592 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843630 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682632 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843636 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843629 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682564 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843601 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682571 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843584 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843639 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843638 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682585 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843566 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843668 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682588 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682581 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682612 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843602 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682605 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682599 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682587 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682539 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682593 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843619 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843664 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843623 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682590 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682625 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843569 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843612 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682534 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682621 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682611 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23843578 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03682598 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 770778 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P379111 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 18137 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 13148 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P117744 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P117744 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 45619 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3747204 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P379111 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P117744 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P117744 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P01154681 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P379111 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P24256 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 112154 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 86097 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01154692 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01154681 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P117744 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01154700 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16986408 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16986421 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16986406 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P497564 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P19511 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P15040334 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8103908 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8103907 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8103910 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8103909 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8103906 | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0400012603.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012603.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012603.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012603.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012603.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012603.hg.1 | ENST00000511465.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012603.hg.1 | ENST00000511465.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012603.hg.1 | NR_040108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012603.hg.1 | NR_040108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012603.hg.1 | NR_040108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012603.hg.1 | NR_040108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04024557.hg.1 | TC04001803.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04024557.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04024557.hg.1 | NR_040108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04024556.hg.1 | TC04001803.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04024556.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04024556.hg.1 | NR_040108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04024553.hg.1 | TC04001803.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04024553.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04024553.hg.1 | NR_040108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04024554.hg.1 | TC04001803.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04024554.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04024554.hg.1 | NR_040108.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400186199.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400186199.hg.1 | ENST00000511465.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400186199.hg.1 | NR_040108.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400186199.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400186199.hg.1 | ENST00000511465.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400186199.hg.1 | NR_040108.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400186199.hg.1 | TC0400012603.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400012603.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012603.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012603.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012603.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012603.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012603.hg.1 | ENST00000511465.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012603.hg.1 | ENST00000511465.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012603.hg.1 | NR_040108.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012603.hg.1 | NR_040108.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012603.hg.1 | NR_040108.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012603.hg.1 | NR_040108.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400186205.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400186205.hg.1 | NR_040108.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400186205.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400186205.hg.1 | NR_040108.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400186205.hg.1 | TC0400012603.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400186198.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400186198.hg.1 | NR_040108.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400186198.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400186198.hg.1 | NR_040108.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400186198.hg.1 | TC0400012603.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400186206.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400186206.hg.1 | NR_040108.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400186206.hg.1 | ENST00000317596.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400186206.hg.1 | NR_040108.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400186206.hg.1 | TC0400012603.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 1554732_at | - | - |
Human Genome U133 Plus 2.0 Array | 1554733_at | - | - |
Human Genome U133 Plus 2.0 Array | 1554732_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1554733_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554732_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554733_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554732_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554732_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554732_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554733_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554733_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554733_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554733_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554733_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554733_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554732_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554732_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554733_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554733_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554733_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554732_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554733_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554732_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554732_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554732_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554732_at10 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A025064 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A025064 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 224007 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3283458 | - | - |
