| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000369718.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000369718.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000369718.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000369718.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000369718.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000369718.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000369718.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000369718.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000464591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000464591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000464591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000464591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000464591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000464591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
ENST00000464591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_001300834.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_001300834.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_001300834.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_001300834.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_001300834.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_001300834.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_001300834.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_001300834.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_001300834.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_001300835.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_001300835.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_001300835.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_001300835.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_001300835.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_001300835.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_001300835.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_001300835.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_001300835.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009417.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000369718.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000369718.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000369718.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000369718.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000369718.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000369718.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000369718.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000369718.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000464591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000464591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000464591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000464591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000464591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000464591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
ENST00000464591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_001300834.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_001300834.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_001300834.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_001300834.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_001300834.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_001300834.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_001300834.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_001300834.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_001300834.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_001300835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_001300835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_001300835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_001300835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_001300835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_001300835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_001300835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_001300835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_001300835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009417.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100167787.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100167787.hg.1 |
ENST00000369718.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100167787.hg.1 |
ENST00000464591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100167787.hg.1 |
NM_001300834.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100167787.hg.1 |
NM_001300835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100167787.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100167787.hg.1 |
ENST00000343534.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100167787.hg.1 |
ENST00000369718.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100167787.hg.1 |
ENST00000464591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100167787.hg.1 |
NM_001300834.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100167787.hg.1 |
NM_001300835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100167787.hg.1 |
NM_174896.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100167787.hg.1 |
TC0100009417.hg.1 |
Clariom D TC |