Assay Details
| Gene Symbol: ANKRD37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181726.3 | NP_859077.1 | 1 - 2 | 268 | 65 | |
| XM_017008176.1 | XP_016863665.1 | 1 - 2 | 113 | 65 | ||
| GenBank mRNA | AK303243.1 | - | 1 - 2 | 143 | 65 | |
| AY296056.1 | - | 1 - 2 | 269 | 65 | ||
| BC088376.1 | - | 1 - 2 | 214 | 30386201 | 65 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000905.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK303243.1,AY296056.1,BC088376.1,NM_181726.3,XM_017008176.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000506424.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000506424.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000506424.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000506424.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000506424.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000506424.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000507479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000507479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000507479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000507479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000507753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000507753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000507753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000507753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000511311.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000511311.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000511311.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000511311.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000511311.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009525.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012363.hg.1 | TC04000905.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012363.hg.1 | ENST00000335174.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012363.hg.1 | ENST00000507479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012363.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012358.hg.1 | TC04000905.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012358.hg.1 | ENST00000335174.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012358.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012359.hg.1 | TC04000905.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012359.hg.1 | ENST00000335174.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012359.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012362.hg.1 | TC04000905.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012362.hg.1 | ENST00000335174.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012362.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400165219.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165219.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400165219.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165219.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400165219.hg.1 | TC0400009525.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400165224.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165224.hg.1 | ENST00000506424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165224.hg.1 | ENST00000507753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165224.hg.1 | ENST00000511311.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165224.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165224.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400165224.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165224.hg.1 | ENST00000506424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165224.hg.1 | ENST00000507753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165224.hg.1 | ENST00000511311.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165224.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165224.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400165224.hg.1 | TC0400009525.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400165225.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165225.hg.1 | ENST00000506424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165225.hg.1 | ENST00000507479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165225.hg.1 | ENST00000507753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165225.hg.1 | ENST00000511311.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165225.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165225.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400165225.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165225.hg.1 | ENST00000506424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165225.hg.1 | ENST00000507479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165225.hg.1 | ENST00000507753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165225.hg.1 | ENST00000511311.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165225.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165225.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400165225.hg.1 | TC0400009525.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000506424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000506424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000506424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000506424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000506424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000506424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000507479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000507479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000507479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000507479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000507753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000507753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000507753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000507753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000511311.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000511311.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000511311.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000511311.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000511311.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009525.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400165223.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165223.hg.1 | ENST00000506424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165223.hg.1 | ENST00000507753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165223.hg.1 | ENST00000511311.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165223.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165223.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400165223.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165223.hg.1 | ENST00000506424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165223.hg.1 | ENST00000507753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165223.hg.1 | ENST00000511311.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165223.hg.1 | ENST00000511393.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165223.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400165223.hg.1 | TC0400009525.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400165220.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165220.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400165220.hg.1 | ENST00000335174.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400165220.hg.1 | NM_181726.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400165220.hg.1 | TC0400009525.hg.1 | Clariom D TC |

ANKRD37
Genomic Map