Assay Details
| Gene Symbol: SPC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317032.1 | NP_001303961.1 | 3 - 4 | 444 | 90 | |
| NM_001317033.1 | NP_001303962.1 | 3 - 4 | 557 | 90 | ||
| NM_182513.3 | NP_872319.1 | 3 - 4 | 444 | 90 | ||
| XM_005259753.3 | XP_005259810.1 | 3 - 4 | 491 | 90 | ||
| GenBank mRNA | AK075287.1 | - | 3 - 4 | 414 | 90 | |
| AK303157.1 | - | 3 - 4 | 301 | 90 | ||
| AK303266.1 | - | 3 - 4 | 552 | 90 | ||
| AY456387.1 | - | 3 - 4 | 408 | 90 | ||
| BC105037.1 | - | 3 - 4 | 440 | 8144040 | 90 | |
| BC105039.1 | - | 3 - 4 | 440 | 8144042 | 90 | |
| BQ061822.1 | - | 3 - 4 | 401 | 90 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001177.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19016354.hg.1 | ENST00000423327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19016354.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19016354.hg.1 | TC19001177.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19016353.hg.1 | ENST00000423327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19016353.hg.1 | ENST00000429831.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19016353.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19016353.hg.1 | TC19001177.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19016352.hg.1 | ENST00000423327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19016352.hg.1 | ENST00000429831.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19016352.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19016352.hg.1 | TC19001177.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000585486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000585486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000585486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000585486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000585486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000585486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000585567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000585567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000585567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000585567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000585567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000585567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000586708.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000586708.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000586708.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000586708.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000586708.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000591396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000591396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000591396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000591396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000591396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000591396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000591396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000592967.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000592967.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000592967.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000592967.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000592967.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | ENST00000592967.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009668.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000585486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000585486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000585486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000585486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000585486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000585486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000585567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000585567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000585567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000585567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000585567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000585567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000586708.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000586708.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000586708.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000586708.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000586708.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000591396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000591396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000591396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000591396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000591396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000591396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000591396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000592967.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000592967.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000592967.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000592967.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000592967.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | ENST00000592967.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009668.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900178270.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178270.hg.1 | ENST00000585486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178270.hg.1 | ENST00000585567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178270.hg.1 | ENST00000586708.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178270.hg.1 | ENST00000591396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178270.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178270.hg.1 | ENST00000592967.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178270.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900178270.hg.1 | TC1900009668.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900178270.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178270.hg.1 | ENST00000585486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178270.hg.1 | ENST00000585567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178270.hg.1 | ENST00000586708.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178270.hg.1 | ENST00000591396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178270.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178270.hg.1 | ENST00000592967.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178270.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900178269.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178269.hg.1 | ENST00000585486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178269.hg.1 | ENST00000586708.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178269.hg.1 | ENST00000591396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178269.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178269.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900178269.hg.1 | TC1900009668.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900178269.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178269.hg.1 | ENST00000585486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178269.hg.1 | ENST00000586708.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178269.hg.1 | ENST00000591396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178269.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178269.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900178271.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178271.hg.1 | ENST00000585486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178271.hg.1 | ENST00000585567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178271.hg.1 | ENST00000591396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178271.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178271.hg.1 | ENST00000592967.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178271.hg.1 | NM_182513.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900178271.hg.1 | TC1900009668.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900178271.hg.1 | ENST00000423327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178271.hg.1 | ENST00000585486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178271.hg.1 | ENST00000585567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178271.hg.1 | ENST00000591396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178271.hg.1 | ENST00000592540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178271.hg.1 | ENST00000592967.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900178271.hg.1 | NM_182513.3 | Refseq |

SPC24
Genomic Map