Assay Details
| Gene Symbol: BPIFB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182519.2 | NP_872325.2 | 6 - 7 | 1005 | 85 | |
| GenBank mRNA | AF549190.1 | - | 5 - 6 | 873 | 85 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000223.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000445356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000445356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000445356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000445356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000445356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000445356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000445356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000494121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000494121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000494121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | ENST00000494121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002788.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002788.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002788.hg.1 | TC20000223.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002789.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002789.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002789.hg.1 | TC20000223.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000445356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000445356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000445356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000445356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000445356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000445356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000445356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000494121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000494121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000494121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | ENST00000494121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007142.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000148915.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148915.hg.1 | ENST00000445356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148915.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000148915.hg.1 | TC2000007142.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000148915.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148915.hg.1 | ENST00000445356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148915.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000148914.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148914.hg.1 | ENST00000445356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148914.hg.1 | NM_182519.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000148914.hg.1 | TC2000007142.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000148914.hg.1 | ENST00000375483.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148914.hg.1 | ENST00000445356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148914.hg.1 | NM_182519.2 | Refseq |

BPIFB4
Genomic Map