Assay Details
| Gene Symbol: LOC284241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136643.1 | 2 - 3 | 520 | 53 | ||
| GenBank mRNA | AK098581.1 | - | 2 - 3 | 520 | 53 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847419 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847462 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451505 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847405 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847497 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451542 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847467 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847400 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847518 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847466 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451570 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847469 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847401 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451563 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847409 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451520 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451545 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847525 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847516 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847431 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451618 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451556 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451611 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847416 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847392 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451560 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847478 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451589 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451487 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847411 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847444 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847436 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451475 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847420 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451582 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847520 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847465 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847398 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847448 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847428 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847505 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847447 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451509 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847395 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847424 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847397 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451601 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847425 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847407 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847412 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451559 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451602 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451491 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847501 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451540 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847415 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847429 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847471 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451499 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451535 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451569 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847423 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847393 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451608 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847438 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847484 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451523 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17089184 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847450 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847390 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847480 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847519 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451598 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847414 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451544 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451480 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847506 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847527 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31451580 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847509 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847485 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847402 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847513 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10847408 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 15123 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P135904 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P135904 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P60585 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P186141 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P20933444 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P135904 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10847543 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10847534 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10847550 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10847535 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10847531 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10847546 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10847551 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10847537 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10847532 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10847545 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10847547 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10847554 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10847553 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10847541 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10847539 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089093 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089176 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089095 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089094 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847499 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089152 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089178 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847430 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089101 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089179 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089099 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847375 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847498 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089096 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847429 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847376 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089170 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847439 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847553 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089219 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847438 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847440 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089102 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089103 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089098 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089222 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089175 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847552 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847442 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847374 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847433 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089171 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847551 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847432 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089220 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847431 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847554 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089174 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089181 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089180 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089172 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847428 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089097 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089204 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847436 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089221 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847441 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847437 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847500 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089177 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089100 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089173 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847330 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10847497 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17089218 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P41112990 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P20933488 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P20933444 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P20933498 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P135904 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P186141 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 119511 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 57448 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 31915 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 27210 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 83522 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20933452 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20933456 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20933480 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20933488 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P41112951 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20933473 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P135904 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20933467 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20933498 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P41112957 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P41113005 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P41112990 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P12965061 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20933444 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P12965440 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P21288332 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8023912 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8023913 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8023911 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8023910 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009190.hg.1 | ENST00000317008.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009190.hg.1 | ENST00000317008.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009190.hg.1 | ENST00000317008.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009190.hg.1 | ENST00000317008.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009190.hg.1 | ENST00000317008.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009190.hg.1 | ENST00000317008.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009190.hg.1 | ENST00000317008.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009190.hg.1 | ENST00000317008.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009190.hg.1 | ENST00000317008.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009190.hg.1 | ENST00000317008.4 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 231952_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 231952_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 688820 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1710829 | AK098581.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1710829 | AK098581.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1710829 | AK098581.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_32698971-S | AK098581.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_7497-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0006620142 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0006620142 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1710829 | AK098581.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1710829 | AK098581.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 2470129 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_7497 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1710829 | AK098581.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1710829 | AK098581.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1710829 | AK098581.1 | human_transcriptome |

LOC284241
Genomic Map