Assay Details
| Gene Symbol: LINC00528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103718.1 | 1 | 1088 | 70 | ||
| GenBank mRNA | AK097861.1 | - | 1 | 1055 | 70 | |
| AW406974.1 | - | 1 | 119 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 691832 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 404953 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P417267 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 7273 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P115473 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P115473 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 56326 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3312985 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P417267 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P115473 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P115473 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P417267 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 72490 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 36866 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03599675 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P879849 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03599675 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8071135 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8071134 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1556072_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006526.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006526.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006526.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006526.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006526.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006526.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006526.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006526.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006526.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006526.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006526.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006526.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006526.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006526.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006526.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006526.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006526.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006526.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006526.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200144629.hg.1 | TC2200006526.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200144629.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144629.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200144629.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144629.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006526.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006526.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006526.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006526.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006526.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006526.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006526.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006526.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006526.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006526.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006526.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006526.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006526.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006526.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006526.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006526.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006526.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006526.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006526.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200144628.hg.1 | TC2200006526.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200144628.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144628.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200144628.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144628.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200144627.hg.1 | TC2200006526.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200144627.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144627.hg.1 | NR_103718.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200144627.hg.1 | ENST00000600723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200144627.hg.1 | NR_103718.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 1556072_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1556072_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556072_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556072_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556072_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556072_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556072_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556072_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556072_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556072_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556072_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556072_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556072_at11 | - | - |
| Human Genome U95 Set | 81664_at | - | - |
| Human Genome U95 Set | 81664_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 229418 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3265879 | AK097861.1 | human_transcriptome |

LINC00528
Genomic Map