Assay Details
| Gene Symbol: ZNF252P-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026974.1 | 1 | 2634 | 108 | ||
| GenBank mRNA | AK092777.1 | - | 1 | 2459 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
zinc finger protein 252, pseudogene |
ZNF252P | ZNF252 | -2.2kb | Sense | Chr. 8 - 144973589 - 145002899 on NCBI Build GRCh38 |
| Downstream | chromosome 8 open reading frame 33 | C8orf33 | 46.8kb | Anti-sense | Chr. 8 - 145052438 - 145056030 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26825717 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26825723 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26825702 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26825736 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06538249 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16151004 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26825707 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06538285 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26825742 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26825708 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26825741 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06538271 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26825703 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06538251 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06538272 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26825690 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26825724 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06538289 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16151016 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06538291 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06538252 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26825725 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06538267 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16151033 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26825696 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16151015 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06538248 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06538265 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06538273 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06538264 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16151012 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26825700 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06538254 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06538263 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26825712 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06538288 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16151005 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26825727 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06538283 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26825704 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 74931 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 32412 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 864953 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 214380 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 210908 | AK125568.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 210908 | AL049686.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 210908 | BC040902.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 210908 | NM_001024661.1 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 210908 | NM_001099854.1 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 210908 | NM_023013.1 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 137086 | AF086169.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 137086 | AK001285.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 137086 | AK026550.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 137086 | AK290761.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 137086 | BC005398.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 137086 | BX537696.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 137086 | CR601244.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 137086 | NM_015391.2 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 360492 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 109070 | AK130659.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 109070 | BC015969.2 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 109070 | CR617464.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 109070 | J03132.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 109070 | M24283.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 109070 | NM_000201.2 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 109070 | X06990.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 369046 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P935906 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P935906 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 223 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 16003 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_14_P119006 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_14_P119006 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P935906 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_32_P935906 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538283 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538242 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538241 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26825724 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26825666 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16151004 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538264 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16151030 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538273 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26825722 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26825726 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16151005 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26825723 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26825712 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538281 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538269 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26825714 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538249 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538267 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538238 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538245 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538252 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538272 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538247 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538263 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26825727 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538271 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538236 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538233 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538251 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538237 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26825722 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538241 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538269 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26825723 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26825714 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538267 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16151004 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538271 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538251 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538264 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26825724 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16151030 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16151005 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538249 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26825666 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538242 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538247 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538237 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538272 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538233 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538236 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538245 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26825726 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538252 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538281 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26825712 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538238 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538283 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538273 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06538263 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26825727 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P935906 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P119006 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P935906 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 107054 | AF119902.1 | genbank_mrna_human |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 59896 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P119006 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P38621331 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538283 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538251 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538268 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538287 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538272 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538220 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538279 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538210 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16151030 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538290 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538273 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538217 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538269 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538223 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538242 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538262 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538215 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538238 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16151004 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538234 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16151008 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538212 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538226 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538237 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538245 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16151025 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538228 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538240 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538276 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538292 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538249 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538219 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06538264 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P119006 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8148954 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8148952 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8148953 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8148951 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1558790_PM_s_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012250.hg.1 | ENST00000525753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012250.hg.1 | ENST00000525753.1 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 1558790_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1558790_PM_s_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558790_s_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558790_s_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558790_s_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558790_s_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558790_s_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558790_s_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558790_s_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558790_s_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558790_s_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558790_s_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558790_s_at8 | - | - |
| Human Genome U95 Set | 89119_PM_at | - | - |
| Human Genome U95 Set | 89119_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037033 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037033 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 230675 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1705085 | AK092777.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1705085 | BC046439.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1705085 | NR_026974.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1705085 | AK092777.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1705085 | BC046439.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1705085 | NR_026974.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1705085 | AK092777.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1705085 | BC046439.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1705085 | NR_026974.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1705085 | AK092777.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1705085 | BC046439.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1705085 | NR_026974.1 | human_transcriptome |

ZNF252P-AS1
Genomic Map