Assay Details
| Gene Symbol: LINC00174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026873.1 | 5 | 2977 | 85 | ||
| GenBank mRNA | AK091213.1 | - | 1 | 1635 | 85 | |
| AK092625.1 | - | 1 | 754 | 85 | ||
| AK124597.1 | - | 1 | 2098 | 85 | ||
| BC020399.1 | - | 5 | 2979 | 85 | ||
| BC093655.1 | - | 1 | 114 | 85 | ||
| BC093657.1 | - | 1 | 114 | 85 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 126121 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 110369 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 16615 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 14891 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 597488 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 259692 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 51316 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 68291 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 410905 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 897214 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 840737 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 771731 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 403373 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 214732 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 107901 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 135975 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 22138 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 112887 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 243444 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 235251 | XM_089866.3 | refseq_mrna_human_XM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 235251 | XM_114987.3 | refseq_mrna_human_XM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 235251 | XM_497258.1 | refseq_mrna_human_XM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 235251 | XM_939622.1 | refseq_mrna_human_XM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 235251 | XM_939626.1 | refseq_mrna_human_XM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 235251 | XM_940443.1 | refseq_mrna_human_XM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 402264 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 215111 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 112156 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 279681 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 212344 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 392219 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | AF083246.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | AF110323.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | AK001218.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | AK001260.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | BC001570.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | BC003056.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | BC008453.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | BC009597.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | BC051898.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | BC107757.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR596038.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR596875.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR597590.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR598771.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR602535.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR603998.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR605115.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR605773.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR605808.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR606506.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR608637.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR611204.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR613733.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR616185.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR617428.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR617662.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR618505.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR618517.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR619763.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR620073.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR621525.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR622891.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR622998.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | CR623406.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168879 | NM_014038.1 | refseq_mrna_human_NM |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P397543 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 35668 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 43462 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 5962 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P16112291 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P16110095 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P32024958 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P397543 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P397543 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25749109 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05511244 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15961669 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05511242 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25749131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15961665 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15961676 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15961655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15961666 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05511243 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25749114 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15961676 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15961655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05511243 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05511244 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15961665 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25749114 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25749131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15961669 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25749109 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15961666 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05511242 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P97496 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P397543 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P121895 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P202399 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05511230 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05511211 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05511216 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05511237 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05511228 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05511245 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05511232 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15961594 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05511221 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05511239 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05511215 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05511218 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05511234 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05511243 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15961591 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05511224 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05511242 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15961589 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P121895 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P202399 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8139874 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8139878 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8139876 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8139875 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8139877 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8139873 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 235397_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 235397_at | - | - |
| Human Genome U133 Plus 2.0 Array | 235397_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235397_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235397_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235397_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235397_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235397_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235397_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235397_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235397_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235397_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235397_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235397_at7 | - | - |
| Human Genome U95 Set | 51800_PM_at | - | - |
| Human Genome U95 Set | 51800_at | - | - |
| Human Genome U95 Set | 76058_PM_at | - | - |
| Human Genome U95 Set | 76058_at | - | - |
| Human Genome U95 Set | 74405_at | - | - |
| Human Genome U95 Set | 74405_PM_at | - | - |
| Human Genome U133 Set | 235397_at | - | - |
| Human Genome U133 Set | 235397_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A032944 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A032944 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 114374 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1733103 | BQ645787.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1733103 | AK092625.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1733103 | AK124597.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1733103 | AW008847.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1733103 | NR_026873.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1733103 | BQ645787.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1733103 | AK092625.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1733103 | AK124597.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1733103 | AW008847.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1733103 | NR_026873.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1733103 | BQ645787.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1733103 | AK092625.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1733103 | AK124597.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1733103 | AW008847.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1733103 | NR_026873.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1733103 | BQ645787.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1733103 | AK092625.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1733103 | AK124597.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1733103 | AW008847.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1733103 | NR_026873.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1733103 | BQ645787.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1733103 | AK092625.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1733103 | AK124597.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1733103 | AW008847.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1733103 | NR_026873.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1733103 | BQ645787.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1733103 | AK092625.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1733103 | AK124597.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1733103 | AW008847.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1733103 | NR_026873.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1733103 | BQ645787.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1733103 | AK092625.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1733103 | AK124597.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1733103 | AW008847.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1733103 | NR_026873.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1733103 | BQ645787.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1733103 | AK092625.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1733103 | AK124597.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1733103 | AW008847.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1733103 | NR_026873.1 | human_transcriptome |

LINC00174
Genomic Map