Assay Details
| Gene Symbol: LOC642852 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026943.1 | 3 | 4182 | 110 | ||
| Gene Symbol: LINC00205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF426264.1 | - | 2 | 110 | ||
| Gene Symbol: LOC107985476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017028526.1 | XP_016884015.1 | 1 | 110 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000243.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P83075 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 38835 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 57656 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P83075 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 115598 | AK097707.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 115598 | NM_145205.3 | refseq_mrna_human_NM |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 17012 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P13944014 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P124118 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | XR_040463_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | XR_040464_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | XR_040465_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8070895 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8070892 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8070894 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8070893 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8070891 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1552819_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11753140_at | - | - |
| Affymetrix Human Genome U219 Array | 11753140_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002874.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002874.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002874.hg.1 | TC21000243.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000400362.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000400362.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000400362.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000400362.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000400362.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002876.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002876.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002876.hg.1 | TC21000243.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002875.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002875.hg.1 | TC21000243.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2100150174.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100150174.hg.1 | TC2100007412.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100150174.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100150177.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100150177.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100150177.hg.1 | TC2100007412.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100150177.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100150177.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100150182.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100150182.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100150182.hg.1 | TC2100007412.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100150182.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100150182.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100150184.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100150184.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100150184.hg.1 | TC2100007412.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100150184.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100150184.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000400362.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000400362.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000400362.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000400362.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000400362.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100150175.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100150175.hg.1 | TC2100007412.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100150175.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100150176.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100150176.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100150176.hg.1 | TC2100007412.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100150176.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100150176.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100150183.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100150183.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100150183.hg.1 | TC2100007412.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100150183.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100150183.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100150178.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100150178.hg.1 | TC2100007412.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100150178.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100150179.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100150179.hg.1 | TC2100007412.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100150179.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100150180.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100150180.hg.1 | TC2100007412.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100150180.hg.1 | NR_026943.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 226995_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1552819_at | - | - |
| Human Genome U133 Plus 2.0 Array | 226995_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1552819_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 228909_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 228908_PM_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 228908_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 228909_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552819_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226995_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228909_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228909_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226995_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552819_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228909_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228908_s_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226995_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552819_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552819_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228909_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228908_s_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226995_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228909_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228909_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228908_s_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228908_s_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228908_s_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228909_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228908_s_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226995_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228908_s_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552819_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226995_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552819_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228909_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228908_s_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552819_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226995_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552819_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228908_s_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228908_s_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552819_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226995_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226995_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228908_s_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226995_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228909_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228909_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 228909_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552819_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 226995_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552819_at1 | - | - |
| Human Genome U95 Set | 65479_at | - | - |
| Human Genome U95 Set | 65479_PM_at | - | - |
| Human Genome U133 Set | 226995_PM_at | - | - |
| Human Genome U133 Set | 228909_at | - | - |
| Human Genome U133 Set | 226995_at | - | - |
| Human Genome U133 Set | 228908_PM_s_at | - | - |
| Human Genome U133 Set | 228908_s_at | - | - |
| Human Genome U133 Set | 228909_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 217451 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 230800 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1780406 | NR_026943.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1780406 | AF426264.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1780406 | NR_026943.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1780406 | AF426264.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1780406 | NR_026943.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1780406 | AF426264.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1780406 | NR_026943.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1780406 | AF426264.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1780406 | NR_026943.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1780406 | AF426264.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1780406 | NR_026943.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1780406 | AF426264.1 | human_transcriptome |

LOC642852
Genomic Map