Assay Details
| Gene Symbol: RPS14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025070.1 | NP_001020241.1 | 1 - 2 | 260 | 91 | |
| GenBank mRNA | BC020515.1 | - | 1 - 2 | 199 | 3856643 | 90 |
| BM454896.1 | - | 1 - 2 | 229 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001937.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC020515.1,NM_001025070.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ribosomal small subunit assembly
negative regulation of transcription from RNA polymerase II promoter
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
maturation of SSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
rRNA processing
translation
translational initiation
regulation of translation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
erythrocyte differentiation
maturation of SSU-rRNA
RNA binding
structural constituent of ribosome
protein binding
poly(A) RNA binding
translation regulator activity
mRNA 5'-UTR binding
small ribosomal subunit rRNA binding
ribosomal small subunit assembly
negative regulation of transcription from RNA polymerase II promoter
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
maturation of SSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
rRNA processing
translation
translational initiation
regulation of translation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
erythrocyte differentiation
maturation of SSU-rRNA
RNA binding
structural constituent of ribosome
protein binding
poly(A) RNA binding
translation regulator activity
mRNA 5'-UTR binding
small ribosomal subunit rRNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05026881.hg.1 | TC05001937.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026881.hg.1 | ENST00000401695.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026881.hg.1 | ENST00000407193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026881.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026881.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026881.hg.1 | NM_005617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026882.hg.1 | TC05001937.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026882.hg.1 | ENST00000401695.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026882.hg.1 | ENST00000407193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026882.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026882.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026882.hg.1 | NM_005617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026880.hg.1 | TC05001937.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026880.hg.1 | ENST00000407193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026880.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026880.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026880.hg.1 | NM_005617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007173.hg.1 | ENST00000457263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007173.hg.1 | NR_077246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007173.hg.1 | NR_077246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007173.hg.1 | NR_077246.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012149.hg.1 | ENST00000496058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000312037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000312037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000312037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000312037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000312037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000312037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000312037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000407193.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000407193.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000407193.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000407193.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000407193.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000407193.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000407193.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000518139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000518139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000518139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000518139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000518139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000518139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000518139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000519855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000519855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000519855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000521466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000521466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000521466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000521466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000521466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000521466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | ENST00000521466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | NM_005617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | NM_005617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | NM_005617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | NM_005617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | NM_005617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013387.hg.1 | NM_005617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026864.hg.1 | TC05001937.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026864.hg.1 | ENST00000312037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026864.hg.1 | ENST00000401695.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026864.hg.1 | ENST00000407193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026864.hg.1 | ENST00000521466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026864.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026864.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026864.hg.1 | NM_005617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008406.hg.1 | ENST00000478778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026871.hg.1 | TC05001937.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026871.hg.1 | ENST00000312037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026871.hg.1 | ENST00000407193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026871.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026871.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017104.hg.1 | ENST00000451736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500197257.hg.1 | ENST00000312037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500197257.hg.1 | ENST00000407193.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500197257.hg.1 | ENST00000518139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500197257.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500197257.hg.1 | ENST00000521466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500197257.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500197257.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500197257.hg.1 | NM_005617.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500197257.hg.1 | ENST00000312037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500197257.hg.1 | ENST00000407193.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500197257.hg.1 | ENST00000518139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500197257.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500197257.hg.1 | ENST00000521466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500197257.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500197257.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500197257.hg.1 | NM_005617.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500197257.hg.1 | TC0500013387.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012149.hg.1 | ENST00000496058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000312037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000312037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000312037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000312037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000312037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000312037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000312037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000407193.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000407193.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000407193.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000407193.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000407193.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000407193.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000407193.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000518139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000518139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000518139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000518139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000518139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000518139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000518139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000519855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000519855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000519855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000521466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000521466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000521466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000521466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000521466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000521466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | ENST00000521466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | NM_005617.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | NM_005617.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | NM_005617.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | NM_005617.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | NM_005617.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013387.hg.1 | NM_005617.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008406.hg.1 | ENST00000478778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500197263.hg.1 | ENST00000312037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500197263.hg.1 | ENST00000407193.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500197263.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500197263.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500197263.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500197263.hg.1 | ENST00000312037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500197263.hg.1 | ENST00000407193.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500197263.hg.1 | ENST00000519690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500197263.hg.1 | NM_001025070.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500197263.hg.1 | NM_001025071.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500197263.hg.1 | TC0500013387.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100017104.hg.1 | ENST00000451736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007173.hg.1 | ENST00000457263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007173.hg.1 | NR_077246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007173.hg.1 | NR_077246.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007173.hg.1 | NR_077246.1 | Refseq |

RPS14
Genomic Map