Assay Details
| Gene Symbol: IZUMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321865.1 | NP_001308794.1 | 1 - 2 | 307 | 65 | |
| NM_182575.2 | NP_872381.2 | 2 - 3 | 782 | 65 | ||
| XM_005258793.4 | XP_005258850.1 | 2 - 3 | 515 | 65 | ||
| XM_011526811.2 | XP_011525113.1 | 1 - 2 | 1286 | 65 | ||
| XM_011526812.2 | XP_011525114.1 | 2 - 3 | 862 | 65 | ||
| XM_011526813.2 | XP_011525115.1 | 2 - 3 | 711 | 65 | ||
| XM_017026646.1 | XP_016882135.1 | 1 - 2 | 1286 | 65 | ||
| GenBank mRNA | AB195682.1 | - | 2 - 3 | 640 | 65 | |
| AY552609.1 | - | 1 - 2 | 234 | 65 | ||
| BC034769.1 | - | 2 - 3 | 782 | 5167853 | 65 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001694.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001321865.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000595517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000595517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000595517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000595517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000595517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000595517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000595517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000595517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000595517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000599871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000599871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000599871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000599871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000599871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000602105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000602105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | ENST00000602105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023109.hg.1 | ENST00000332955.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023109.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023109.hg.1 | TC19001694.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023110.hg.1 | ENST00000332955.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023110.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023110.hg.1 | TC19001694.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000595517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000595517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000595517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000595517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000595517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000595517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000595517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000595517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000595517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000599871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000599871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000599871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000599871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000599871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000602105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000602105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | ENST00000602105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011095.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900192789.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192789.hg.1 | ENST00000595517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192789.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192789.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192789.hg.1 | ENST00000602105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192789.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900192789.hg.1 | TC1900011095.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900192789.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192789.hg.1 | ENST00000595517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192789.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192789.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192789.hg.1 | ENST00000602105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192789.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900192791.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192791.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192791.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192791.hg.1 | NM_182575.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900192791.hg.1 | TC1900011095.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900192791.hg.1 | ENST00000332955.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192791.hg.1 | ENST00000595937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192791.hg.1 | ENST00000597553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900192791.hg.1 | NM_182575.2 | Refseq |

IZUMO1
Genomic Map