Assay Details
| Gene Symbol: SMYD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052928.2 | NP_443160.2 | 5 - 6 | 1705 | 60 | |
| XM_011523632.2 | XP_011521934.1 | 5 - 6 | 1686 | 60 | ||
| XM_011523633.2 | XP_011521935.1 | 5 - 6 | 1686 | 60 | ||
| XM_017024121.1 | XP_016879610.1 | 6 - 7 | 1950 | 60 | ||
| GenBank mRNA | AK057769.1 | - | 1 - 2 | 137 | 60 | |
| AK092149.1 | - | 1 - 2 | 610 | 60 | ||
| BC035077.2 | - | 5 - 6 | 1624 | 5260399 | 60 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000984.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17013182.hg.1 | ENST00000305513.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17013182.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17013182.hg.1 | TC17000984.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17013181.hg.1 | ENST00000305513.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17013181.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17013181.hg.1 | TC17000984.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000476292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000476292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000476292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000576633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000576633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000576633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000576633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000582190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000582190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000476292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000476292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000476292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000576633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000576633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000576633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000576633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000582190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000582190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700172028.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700172028.hg.1 | ENST00000491788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700172028.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700172028.hg.1 | ENST00000305513.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700172028.hg.1 | ENST00000491788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700172028.hg.1 | NM_052928.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700172028.hg.1 | TC1700009402.hg.1 | Clariom D TC |

SMYD4
Genomic Map