Assay Details
| Gene Symbol: PNKD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015488.4 | NP_056303.3 | 5 - 6 | 666 | 92 | |
| NM_022572.4 | NP_072094.1 | 4 - 5 | 539 | 92 | ||
| XM_017003771.1 | XP_016859260.1 | 5 - 6 | 573 | 92 | ||
| XM_017003772.1 | XP_016859261.1 | 4 - 5 | 564 | 92 | ||
| GenBank mRNA | AB033010.1 | - | 5 - 6 | 507 | 92 | |
| AK289867.1 | - | 5 - 6 | 574 | 92 | ||
| AK309308.1 | - | 369 | 92 | |||
| AY358753.1 | - | 4 - 5 | 459 | 92 | ||
| BC002937.2 | - | 4 - 5 | 450 | 3956630 | 92 | |
| BC036457.2 | - | 5 - 6 | 568 | 4797574 | 92 | |
| BI764334.1 | - | 4 - 5 | 525 | 92 | ||
| BU553179.1 | - | 4 - 5 | 571 | 92 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001297.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
methylglyoxal catabolic process to D-lactate via S-lactoyl-glutathione
regulation of synaptic transmission, dopaminergic
regulation of dopamine metabolic process
negative regulation of neurotransmitter secretion
neuromuscular process controlling posture
hydroxyacylglutathione hydrolase activity
protein binding
metal ion binding
methylglyoxal catabolic process to D-lactate via S-lactoyl-glutathione
regulation of synaptic transmission, dopaminergic
regulation of dopamine metabolic process
negative regulation of neurotransmitter secretion
neuromuscular process controlling posture
hydroxyacylglutathione hydrolase activity
protein binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000248451.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000248451.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000248451.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000248451.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000248451.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000248451.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000248451.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000469689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000469689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000469689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000469689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000469689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000472650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000472650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000472650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000483797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000483797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000483797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000494954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000494954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000494954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | ENST00000622701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_001077399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_001077399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_001077399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_001077399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_001077399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_001077399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_001077399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010791.hg.1 | NR_106868.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019808.hg.1 | TC02001297.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019808.hg.1 | ENST00000258362.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019808.hg.1 | ENST00000273077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019808.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019808.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019808.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176647.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176647.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176647.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176647.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176647.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176647.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176647.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176647.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176647.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176647.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176647.hg.1 | TC0200010791.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000248451.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000248451.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000248451.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000248451.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000248451.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000248451.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000248451.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000258362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000273077.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000436005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000469689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000469689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000469689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000469689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000469689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000472650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000472650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000472650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000483797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000483797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000483797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000494954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000494954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000494954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | ENST00000622701.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_001077399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_001077399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_001077399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_001077399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_001077399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_001077399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_001077399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_015488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NM_022572.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010791.hg.1 | NR_106868.1 | Refseq |

PNKD
Genomic Map